Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs, approximately 10% cross-reactivity with recombinant human IGFBP-1 and less than 1% cross-reactivity with recombinant mouse (rm) IGFBP-2, rmIGFBP-3, rmIGFBP-5, rmIGFBP-6, and rmIGFBP-7 is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Binding protein 25; Binding protein 26; Binding protein 28; hIGFBP 1; hIGFBP1; IBP 1; IBP-1; IGF BP25; IGF-binding protein 1; IGFBP; IGFBP 1; Insulin like growth factor binding protein; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 1 PRECURSOR (IGFBP-1) (IBP-1) (IGF-BINDING PROTEIN 1); Insulin-like growth factor-binding protein 1; PP 12
Gene Aliases: Igfbp-1; Igfbp1
UniProt ID: (Mouse) P47876
Entrez Gene ID: (Mouse) 16006
Molecular Function:
protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support