Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: SKALHVTNIK KWKEPCRIEL YRVVESLAKA QETSGEEISK FYLPNCNKNG FYHSRQCETS MDGEAGLCWC VYPWNGKRIP GSPEIRGDPN CQI
Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.
This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-pregnancy-associated endometrial globulin; amniotic fluid binding protein; Binding protein 25; Binding protein 26; Binding protein 28; binding protein-25; binding protein-26; binding protein-28; growth hormone independent-binding protein; hIGFBP 1; hIGFBP1; IBP 1; IBP-1; IGF BP25; IGF-binding protein 1; IGFBP; IGFBP 1; IGFBP-1; Insulin like growth factor binding protein; insulin-like growth factor binding protein 1; Insulin-like growth factor-binding protein 1; Placental protein 12; PP 12; PP12
Gene Aliases: AFBP; hIGFBP-1; IBP1; IGF-BP25; IGFBP1; PP12
UniProt ID: (Human) P08833
Entrez Gene ID: (Human) 3484
Molecular Function:
protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support