Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: RALERLAASQ SRTHEDLYII PIPNCDRNGN FHPKQCHPAL DGQRGKCWCV
Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%
This gene is a member of the insulin-like growth factor binding protein family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Band 3A; differentially expressed in B16F10 2; IBP-4; IGF-binding protein 4; IGFBP; Insulin like growth factor binding protein; insulin-like growth factor binding protein 4; Insulin-like growth factor-binding protein 4
Gene Aliases: AI875747; BP-4; Deb2; HT29-IGFBP; IBP4; IGFBP-4; IGFBP4
UniProt ID: (Human) P22692, (Mouse) P47879
Entrez Gene ID: (Human) 3487, (Mouse) 16010
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support