Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
In direct ELISAs, no cross-reactivity with recombinant human (rh) IGF-I, rhIGF-II, rhIGFL-1, rhIGFL-4, and recombinant mouse IGFL-3 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
IGFL3 belongs to the insulin-like growth factor (IGF) family of signaling molecules that play critical roles in cellular energy metabolism and in growth and development, especially prenatal growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IGF; IGF-like family member 3; Insulin growth factor-like family member 3; Insulin like growth factor; RPRC483
Gene Aliases: IGFL3; UNQ483; UNQ483/PRO982
UniProt ID: (Human) Q6UXB1
Entrez Gene ID: (Human) 388555
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support