Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive Samples: HepG2, A-549, Mouse brain, Mouse testis, Rat brain
Immunogen sequence: PYNLQVDLLR QSLVHRHPEL EIKSVDGFQG REKEAVILSF VRSNRKGEVG FLAEDRRINV AVTRARRHVA VICDSRTVNN HAFLKTLVEY FTQHGEVRTA FEYLDDIVPE NYSHENSQGS SHAATKPQGP ATSTRTGSQR QEGGQEAAAP ARQGRKKPAG KSLASEAPSQ PSLNGGSPEG V
5' to 3' helicase that unwinds RNA and DNA duplices in an ATP-dependent reaction. Acts as a transcription regulator. Required for the transcriptional activation of the flounder liver-type antifreeze protein gene. Exhibits strong binding specificity to the enhancer element B of the flounder antifreeze protein gene intron. Binds to the insulin II gene RIPE3B enhancer region. May be involved in translation. DNA-binding protein specific to 5'-phosphorylated single-stranded guanine-rich sequence related to the immunoglobulin mu chain switch region. Preferentially binds to the 5'-GGGCT-3' motif. Interacts with tRNA-Tyr. Stimulates the transcription of the human neurotropic virus JCV.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AEP; antifreeze enhancer-binding protein; Antifreeze enhancer-binding protein ortholog; ATP-dependent helicase IGHMBP2; cardiac transcription factor 1; DNA-binding protein SMUBP-2; GF-1; Glial factor 1; Immunoglobulin mu-binding protein 2; immunoglobulin S mu binding protein 2; neuromuscular degeneration; p110 subunit; zinc finger, AN1-type domain 7
Gene Aliases: AEP; CATF1; CMT2S; HCSA; HMN6; IGHMBP2; nmd; RIPE3b1; sma; SMARD1; Smbp-2; SMBP2; SMUBP2; ZFAND7
UniProt ID: (Human) P38935, (Rat) Q9EQN5
Entrez Gene ID: (Human) 3508, (Mouse) 20589, (Rat) 29532
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support