Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
J chain is a small glycopeptide and is structurally unrelated to heavy or light chains, but is synthesized by all plasma cells that secrete polymeric immunoglobulins. J chain is linked to IgA and IgM by disulfide bonds, however, it has been detected in IgG- and IgD-containing cells. J chains are present in a large proportion of the immunoglobulin-positive cells in the germinal centers of the tonsil and lymph node. B cells secrete J chain at an early stage of differentiation with the expression persisting in those cells destined to produce IgA or IgM.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IgJ chain; Immunoglobulin J chain; immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides; J chain; Joining chain of multimeric IgA and IgM
Gene Aliases: IGCJ; IGJ; JCH; JCHAIN
UniProt ID: (Human) P01591
Entrez Gene ID: (Human) 3512
Molecular Function: defense/immunity protein immunoglobulin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support