Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MSEVKSRKKS GPKGAPAAEP GKRSEGGKTP VARSSGGGGW ADPRTCLSLL SLGTCLGLAW FVFQQSEKFA KVENQYQLLK LETNEFQQLQ SKISLISEKL ESTESILQEA TSSMSLMTQF EQEVSNLQDI MHDIQNNEEV LTQRMQSLNE KFQNITDFWK RSLEEMNINT DIFKSEAKHI HSQVTVQINS AEQEIKLLTE RLKDLEDSTL RNIRTVKRQE EEDLLRVEEQ LGSDTKAIEK LEEEQHALFA RDEDLTNKLS DYEPKVEECK THLPTIESAI HSVLRVSQDL IETEKKMEDL TMQMFNMEDD MLKAVSEIME MQKTLEGIQY DNSILKMQNE LDILKEKVHD FIAYSSTGEK GTLKEYNIEN KGIGGDF
IKIP (Inhibitor of nuclear factor kappa-B kinase-interacting protein, IKK-interacting protein) is a single-pass membrane protein that shares a common promoter with APAF1. APAF1 and IKIP are both induced by X irradiation, however, the two gene products are transcribed in different directions. The IKIP gene is believed to be a target for p53 as expression of IKIP has been shown to promote apoptosis. IKIP has four known isoforms, three of which are found traversing the endoplasmic reticulum membrane. IKIP isoform 4 has a deletion of the transmembrane region which leads to a homogenous distribution of the protein within the cell. The IKIP gene products are expressed in vascular endothelial cells, while the isoform 4 has also been detected in lung, kidney, spleen, thymus and skeletal muscle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: I kappa B kinase interacting protein; I kappa-B kinase-interacting protein; IKBKB-interacting protein; IKK interacting protein; IKK-interacting protein; Inhibitor of nuclear factor kappa-B kinase-interacting protein
Gene Aliases: IKBIP; IKIP
UniProt ID: (Human) Q70UQ0
Entrez Gene ID: (Human) 121457
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support