Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Rockland
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This is an IgG preparation of whole rabbit serum purified by DEAE chromatography. In general, this antibody also detects primate IL-1Ra in the same formats using similar dilutions, but not mouse or rabbit IL-1Ra. The antibody does not recognize human IL-1a, IL-1b or IL-1 receptor type I and type II. This antibody is primarily directed against the secreted form of IL-1Ra (also termed sIL-1Ra) but recognizes the native (non-denatured) as well as denatured intracellular form of IL-1Ra (also termed icIL-1Ra). IcIL-1Ra is found in many cells as constitutively expressed protein, particularly epithelial cells and keratinocytes. This antibody does not distinguish between sIL-1Ra and icIL-1Ra in ELISA formats or by immunohistochemistry. When used in immunoblot analysis, it will recognize 17,000 MW IL-1Ra in supernatants released from supernatants or lysates of 2 x 106 endotoxin-stimulated human peripheral blood mononuclear cells (PBMC). PBMC are stimulated for 24 hours with 1% (v/v) human serum plus 10 ng/mL E.coli LPS. However, in lysates of PBMC, icIL-1Ra is also found after 48-72 hours of culture. In monocytic cells, the initial IL-1Ra transcript is for sIL-1Ra which can be found almost exclusively intracellularly.
The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD121A; D2S1473; ICIL-1RA; IL 1 receptor antagonist; il-1 receptor; il-1r; IL-1R-alpha; IL-1RN; IL1 inhibitor; il1 receptor; IL1 receptor antagonist; IL1-RA; IL1R; IL1RN (IL1F3); interleukin-1 receptor; Interleukin-1 receptor antagonist protein; intracellular IL-1 receptor antagonist type II; intracellular interleukin-1 receptor antagonist (icIL-1ra); MGC10430; P80; type II interleukin-1 receptor antagonist
Gene Aliases: DIRA; ICIL-1RA; IL-1ra; IL-1ra3; IL-1RN; IL1F3; IL1RA; IL1RN; IRAP; MVCD4
UniProt ID: (Human) P18510
Entrez Gene ID: (Human) 3557
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support