Product References
Bi-directional neuro-immune dysfunction after chronic experimental brain injury.
Journal of neuroinflammation
Ritzel RM,Li Y,Jiao Y,Doran SJ,Khan N,Henry RJ,Brunner K,Loane DJ,Faden AI,Szeto GL,Wu J
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Fri Apr 05 00:00:00 UTC 2024
Zika Virus Infection Induces Interleukin-1β-Mediated Inflammatory Responses by Macrophages in the Brain of an Adult Mouse Model.
Journal of virology
Jeong GU,Lee S,Kim DY,Lyu J,Yoon GY,Kim KD,Ku KB,Ko J,Kwon YC
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Thu Jun 29 00:00:00 UTC 2023
Inhibition of autophagy in microglia and macrophages exacerbates innate immune responses and worsens brain injury outcomes.
Autophagy
Hegdekar N,Sarkar C,Bustos S,Ritzel RM,Hanscom M,Ravishankar P,Philkana D,Wu J,Loane DJ,Lipinski MM
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Sat Jul 01 00:00:00 UTC 2023
Low-dose adropin stimulates inflammasome activation of macrophage via mitochondrial ROS involved in colorectal cancer progression.
BMC cancer
Jia L,Liao L,Jiang Y,Hu X,Lu G,Xiao W,Gong W,Jia X
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Mon Oct 30 00:00:00 UTC 2023
Microbially-Induced Exosomes from Dendritic Cells Promote Paracrine Immune Senescence: Novel Mechanism of Bone Degenerative Disease in Mice.
Aging and disease
Elsayed R,Elashiry M,Liu Y,Morandini AC,El-Awady A,Elashiry MM,Hamrick M,Cutler CW
12-7114-82 was used in Flow cytometry/Cell sorting to conclude that immune senescence, compounded by advanced age, and accelerated by oral dysbiosis and its induced SASP exosomes, plays a pivotal role in the pathophysiology of experimental periodontitis.
Wed Feb 01 00:00:00 UTC 2023
Brain innate immune response via miRNA-TLR7 sensing in polymicrobial sepsis.
Brain, behavior, and immunity
Zou L,He J,Gu L,Shahror RA,Li Y,Cao T,Wang S,Zhu J,Huang H,Chen F,Fan X,Wu J,Chao W
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Tue Feb 01 00:00:00 UTC 2022
TNF-Like Ligand 1 Aberrance Aggravates Nonalcoholic Steatohepatitis via M1 Macrophage Polarization.
Oxidative medicine and cellular longevity
Luo Y,Guo J,Jia W,Wu M,Yin F,Niu G,Shih DQ,Targan SR,Zhang X
12-7114-82 was used in Flow cytometry/Cell sorting to investigate the role of TNF-like ligand 1 aberrance (TL1A) in nonalcoholic steatohepatitis (NASH).
Fri Mar 04 00:00:00 UTC 2022
TNF-Like Ligand 1 Aberrance Aggravates Nonalcoholic Steatohepatitis via M1 Macrophage Polarization.
Oxidative medicine and cellular longevity
Luo Y,Guo J,Jia W,Wu M,Yin F,Niu G,Shih DQ,Targan SR,Zhang X
12-7114-82 was used in Flow cytometry/Cell sorting to investigate the role of TNF-like ligand 1 aberrance (TL1A) in nonalcoholic steatohepatitis (NASH).
Fri Mar 04 00:00:00 UTC 2022
Smek1 deficiency exacerbates experimental autoimmune encephalomyelitis by activating proinflammatory microglia and suppressing the IDO1-AhR pathway.
Journal of neuroinflammation
Duan RN,Yang CL,Du T,Liu A,Wang AR,Sun WJ,Li X,Li JX,Yan CZ,Liu QJ
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Mon Jun 28 00:00:00 UTC 2021
Smek1 deficiency exacerbates experimental autoimmune encephalomyelitis by activating proinflammatory microglia and suppressing the IDO1-AhR pathway.
Journal of neuroinflammation
Duan RN,Yang CL,Du T,Liu A,Wang AR,Sun WJ,Li X,Li JX,Yan CZ,Liu QJ
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Mon Jun 28 00:00:00 UTC 2021
Smek1 deficiency exacerbates experimental autoimmune encephalomyelitis by activating proinflammatory microglia and suppressing the IDO1-AhR pathway.
Journal of neuroinflammation
Duan RN,Yang CL,Du T,Liu A,Wang AR,Sun WJ,Li X,Li JX,Yan CZ,Liu QJ
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Mon Jun 28 00:00:00 UTC 2021
NLRP3 Inflammasome Activation of Mast Cells by Estrogen via the Nuclear-Initiated Signaling Pathway Contributes to the Development of Endometriosis.
Frontiers in immunology
Guo X,Xu X,Li T,Yu Q,Wang J,Chen Y,Ding S,Zhu L,Zou G,Zhang X
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Tue Jan 18 00:00:00 UTC 2022
MicroRNA-326 attenuates immune escape and prevents metastasis in lung adenocarcinoma by targeting PD-L1 and B7-H3.
Cell death discovery
Shao L,He Q,Wang J,He F,Lin S,Wu L,Gao Y,Ma W,Dong J,Yang X,Li F
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Tue Jun 15 00:00:00 UTC 2021
Fructose reprogrammes glutamine-dependent oxidative metabolism to support LPS-induced inflammation.
Nature communications
Jones N,Blagih J,Zani F,Rees A,Hill DG,Jenkins BJ,Bull CJ,Moreira D,Bantan AIM,Cronin JG,Avancini D,Jones GW,Finlay DK,Vousden KH,Vincent EE,Thornton CA
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Mon Feb 22 00:00:00 UTC 2021
Interleukin-1β Suppresses Gastrin via Primary Cilia and Induces Antral Hyperplasia.
Cellular and molecular gastroenterology and hepatology
Ding L,Sontz EA,Saqui-Salces M,Merchant JL
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Tue Mar 08 00:00:00 UTC 2022
The Intermucosal Connection between the Mouth and Gut in Commensal Pathobiont-Driven Colitis.
Cell
Kitamoto S,Nagao-Kitamoto H,Jiao Y,Gillilland MG,Hayashi A,Imai J,Sugihara K,Miyoshi M,Brazil JC,Kuffa P,Hill BD,Rizvi SM,Wen F,Bishu S,Inohara N,Eaton KA,Nusrat A,Lei YL,Giannobile WV,Kamada N
12-7114-82 was used in Flow Cytometry to conclude that oral inflammation, such as periodontitis, exacerbates gut inflammation by supplying the gut with both colitogenic pathobionts and pathogenic T cells.
Thu Jul 23 00:00:00 UTC 2020
T cells instruct myeloid cells to produce inflammasome-independent IL-1β and cause autoimmunity.
Nature immunology
Jain A,Irizarry-Caro RA,McDaniel MM,Chawla AS,Carroll KR,Overcast GR,Philip NH,Oberst A,Chervonsky AV,Katz JD,Pasare C
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Wed Jan 01 00:00:00 UTC 2020
Endogenous oxidized phospholipids reprogram cellular metabolism and boost hyperinflammation.
Nature immunology
Di Gioia M,Spreafico R,Springstead JR,Mendelson MM,Joehanes R,Levy D,Zanoni I
12-7114 was used in Flow Cytometry to report that the self-encoded oxidized phospholipid oxPAPC alters the metabolism of macrophages exposed to lipopolysaccharide.
Wed Jan 01 00:00:00 UTC 2020
Hippocampal interleukin-33 mediates neuroinflammation-induced cognitive impairments.
Journal of neuroinflammation
Reverchon F,de Concini V,Larrigaldie V,Benmerzoug S,Briault S,Togbé D,Ryffel B,Quesniaux VFJ,Menuet A
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Fri Sep 11 00:00:00 UTC 2020
Hippocampal interleukin-33 mediates neuroinflammation-induced cognitive impairments.
Journal of neuroinflammation
Reverchon F,de Concini V,Larrigaldie V,Benmerzoug S,Briault S,Togbé D,Ryffel B,Quesniaux VFJ,Menuet A
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Fri Sep 11 00:00:00 UTC 2020
Triacylglycerol synthesis enhances macrophage inflammatory function.
Nature communications
Castoldi A,Monteiro LB,van Teijlingen Bakker N,Sanin DE,Rana N,Corrado M,Cameron AM,Hässler F,Matsushita M,Caputa G,Klein Geltink RI,Büscher J,Edwards-Hicks J,Pearce EL,Pearce EJ
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Fri Aug 14 00:00:00 UTC 2020
Mannose receptor high, M2 dermal macrophages mediate nonhealing Leishmania major infection in a Th1 immune environment.
The Journal of experimental medicine
Lee SH,Charmoy M,Romano A,Paun A,Chaves MM,Cope FO,Ralph DA,Sacks DL
12-7114 was used in Flow cytometry/Cell sorting to study the origin and functional specialisation of dermal macrophages in cutaneous infections.
Tue Jan 02 00:00:00 UTC 2018
1,25-Dihydroxyvitamin D suppresses M1 macrophages and promotes M2 differentiation at bone injury sites.
JCI insight
Wasnik S,Rundle CH,Baylink DJ,Yazdi MS,Carreon EE,Xu Y,Qin X,Lau KW,Tang X
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Thu Sep 06 00:00:00 UTC 2018
Dicer Deficiency Differentially Impacts Microglia of the Developing and Adult Brain.
Immunity
Varol D,Mildner A,Blank T,Shemer A,Barashi N,Yona S,David E,Boura-Halfon S,Segal-Hayoun Y,Chappell-Maor L,Keren-Shaul H,Leshkowitz D,Hornstein E,Fuhrmann M,Amit I,Maggio N,Prinz M,Jung S
12-7114 was used in Flow cytometry/Cell sorting to examine the impact of post-transcriptional control mediated by microRNAs on microglial performance during development and adulthood.
Tue Jun 20 00:00:00 UTC 2017
Cholesterol Accumulation in Dendritic Cells Links the Inflammasome to Acquired Immunity.
Cell metabolism
Westerterp M,Gautier EL,Ganda A,Molusky MM,Wang W,Fotakis P,Wang N,Randolph GJ,D'Agati VD,Yvan-Charvet L,Tall AR
12-7114 was used in Flow cytometry/Cell sorting to identify an essential role of DC cholesterol efflux pathways in maintaining immune tolerance.
Tue Jun 06 00:00:00 UTC 2017
Notch Regulates Macrophage-Mediated Inflammation in Diabetic Wound Healing.
Frontiers in immunology
Kimball AS,Joshi AD,Boniakowski AE,Schaller M,Chung J,Allen R,Bermick J,Carson WF,Henke PK,Maillard I,Kunkel SL,Gallagher KA
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Wed Nov 20 00:00:00 UTC 2019
Depletion of microglia exacerbates postischemic inflammation and brain injury.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Jin WN,Shi SX,Li Z,Li M,Wood K,Gonzales RJ,Liu Q
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Thu Jun 01 00:00:00 UTC 2017
The amino acid sensor GCN2 controls gut inflammation by inhibiting inflammasome activation.
Nature
Ravindran R,Loebbermann J,Nakaya HI,Khan N,Ma H,Gama L,Machiah DK,Lawson B,Hakimpour P,Wang YC,Li S,Sharma P,Kaufman RJ,Martinez J,Pulendran B
12-7114 was used in Flow cytometry/Cell sorting to reveal a mechanism that couples amino acid sensing with control of intestinal inflammation via GCN2.
Thu Mar 24 00:00:00 UTC 2016
Neutrophils Oppose Uterine Epithelial Carcinogenesis via Debridement of Hypoxic Tumor Cells.
Cancer cell
Blaisdell A,Crequer A,Columbus D,Daikoku T,Mittal K,Dey SK,Erlebacher A
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Mon Dec 14 00:00:00 UTC 2015
Blood-brain barrier dysfunction developed during normal aging is associated with inflammation and loss of tight junctions but not with leukocyte recruitment.
Immunity & ageing : I & A
Elahy M,Jackaman C,Mamo JC,Lam V,Dhaliwal SS,Giles C,Nelson D,Takechi R
Wed Mar 18 00:00:00 UTC 2015
Differential ASC requirements reveal a key role for neutrophils and a noncanonical IL-1β response to Pseudomonas aeruginosa.
American journal of physiology. Lung cellular and molecular physiology
Patankar YR,Mabaera R,Berwin B
12-7114 was used in Flow cytometry/Cell sorting to demonstrate the physiological contribution of neutrophils as a source of IL-1β in response to P. aeruginosa infection.
Thu Oct 15 00:00:00 UTC 2015
Interleukin-17-producing innate lymphoid cells and the NLRP3 inflammasome facilitate obesity-associated airway hyperreactivity.
Nature medicine
Kim HY,Lee HJ,Chang YJ,Pichavant M,Shore SA,Fitzgerald KA,Iwakura Y,Israel E,Bolger K,Faul J,DeKruyff RH,Umetsu DT
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Wed Jan 01 00:00:00 UTC 2014
The frustrated host response to Legionella pneumophila is bypassed by MyD88-dependent translation of pro-inflammatory cytokines.
PLoS pathogens
Asrat S,Dugan AS,Isberg RR
12-7114 was used in Flow cytometry/Cell sorting to show how the protein translation inhibition induced by L. pneumophila is bypassed by MyD88, allowing pro-inflammatory cytokine translation.
Tue Jul 01 00:00:00 UTC 2014
Acidosis potentiates the host proinflammatory interleukin-1β response to Pseudomonas aeruginosa infection.
Infection and immunity
Torres IM,Patankar YR,Shabaneh TB,Dolben E,Hogan DA,Leib DA,Berwin BL
12-7114 was used in Flow cytometry/Cell sorting to show that extracellular pH governs the host inflammatory response against P. aeruginosa via synergistic mechanisms that modulate cytokine release.
Sat Nov 01 00:00:00 UTC 2014
Cell-selective inhibition of NF-κB signaling improves therapeutic index in a melanoma chemotherapy model.
Cancer discovery
Enzler T,Sano Y,Choo MK,Cottam HB,Karin M,Tsao H,Park JM
Published figure using IL-1 beta (Pro-form) monoclonal antibody (Product # 12-7114-82) in Flow Cytometry
Tue Nov 01 00:00:00 UTC 2011