Product References
Myeloid cell-specific topoisomerase 1 inhibition using DNA origami mitigates neuroinflammation.
EMBO reports
Zhu K,Wang Y,Sarlus H,Geng K,Nutma E,Sun J,Kung SY,Bay C,Han J,Min JH,Benito-Cuesta I,Lund H,Amor S,Wang J,Zhang XM,Kutter C,Guerreiro-Cacais AO,Högberg B,Harris RA
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Tue Jul 05 00:00:00 EDT 2022
RORγt phosphorylation protects against T cell-mediated inflammation.
Cell reports
Ma S,Patel SA,Abe Y,Chen N,Patel PR,Cho BS,Abbasi N,Zeng S,Schnabl B,Chang JT,Huang WJM
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Tue Mar 15 00:00:00 EDT 2022
Gut epithelial IL-27 confers intestinal immunity through the induction of intraepithelial lymphocytes.
The Journal of experimental medicine
Lin CH,Chen MC,Lin LL,Christian DA,Min B,Hunter CA,Lu LF
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Mon Nov 01 00:00:00 EDT 2021
Irradiated whole-cell vaccine suppresses hepatocellular carcinoma growth in mice via Th9 cells.
Oncology letters
Chen J,Ding Y,Huang F,Lan R,Wang Z,Huang W,Chen R,Wu B,Fu L,Yang Y,Liu J,Hong J,Zhang W,Zhang L
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Sat May 01 00:00:00 EDT 2021
Milk-derived extracellular vesicles alleviate ulcerative colitis by regulating the gut immunity and reshaping the gut microbiota.
Theranostics
Tong L,Hao H,Zhang Z,Lv Y,Liang X,Liu Q,Liu T,Gong P,Zhang L,Cao F,Pastorin G,Lee CN,Chen X,Wang JW,Yi H
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Fri Jan 28 00:00:00 EST 2022
Characterisation of Macrophage Polarisation in Mice Infected with Ninoa Strain of Trypanosoma cruzi.
Pathogens (Basel, Switzerland)
Medina-Buelvas DM,Rodríguez-Sosa M,Vega L
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Sat Nov 06 00:00:00 EDT 2021
Retinoic Acid Induces Functionally Suppressive Foxp3+RORγt+ T Cells In Vitro.
Frontiers in immunology
Martínez-Blanco M,Lozano-Ojalvo D,Pérez-Rodríguez L,Benedé S,Molina E,López-Fandiño R
11-7101-41 was used in Flow Cytometry to show that under the influence of IL-2 and TGF-β, suppressive Foxp3+RORγt+ T cells that express and secrete IL-17 can be produced in vitro and RA further contributes to stabilize this phenotype.
Thu Oct 21 00:00:00 EDT 2021
Local Sustained GM-CSF Delivery by Genetically Engineered Encapsulated Cells Enhanced Both Cellular and Humoral SARS-CoV-2 Spike-Specific Immune Response in an Experimental Murine Spike DNA Vaccination Model.
Vaccines
Vernet R,Charrier E,Cosset E,Fièvre S,Tomasello U,Grogg J,Mach N
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Mon May 10 00:00:00 EDT 2021
Blockade of TLR2 and TLR4 Attenuates Inflammatory Response and Parasite Load in Cutaneous Leishmaniasis.
Frontiers in immunology
Carneiro PP,Dórea AS,Oliveira WN,Guimarães LH,Brodskyn C,Carvalho EM,Bacellar O
11-7101-41 was used in Flow Cytometry to show that reduction in oxidative burst, IL-1β, TNF and CXCL9 production were observed.
Mon Dec 13 00:00:00 EST 2021
Tregitopes regulate the tolerogenic immune response and decrease the foetal death rate in abortion-prone mouse matings.
Scientific reports
Kedzierska AE,Lorek D,Slawek A,Chelmonska-Soyta A
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Mon Jun 29 00:00:00 EDT 2020
Memory CD4+ T cells enhance B-cell responses to drifting influenza immunization.
European journal of immunology
Gage E,Van Hoeven N,Dubois Cauwelaert N,Larsen SE,Erasmus J,Orr MT,Coler RN
11-7101-41 was used in Flow Cytometry to show that MHC Class II haplotype and T-cell reactivity influences subsequent hemagglutination inhibitory antibody responses.
Fri Feb 01 00:00:00 EST 2019
c-Maf controls immune responses by regulating disease-specific gene networks and repressing IL-2 in CD4+ T cells.
Nature immunology
Gabryšová L,Alvarez-Martinez M,Luisier R,Cox LS,Sodenkamp J,Hosking C,Pérez-Mazliah D,Whicher C,Kannan Y,Potempa K,Wu X,Bhaw L,Wende H,Sieweke MH,Elgar G,Wilson M,Briscoe J,Metzis V,Langhorne J,Luscombe NM,O'Garra A
11-7101 was used in Flow cytometry/Cell sorting to study the role of c-Maf as a regulator of the expression of cytokine-encoding genes.
Tue May 01 00:00:00 EDT 2018
Excessive expression of miR-27 impairs Treg-mediated immunological tolerance.
The Journal of clinical investigation
Cruz LO,Hashemifar SS,Wu CJ,Cho S,Nguyen DT,Lin LL,Khan AA,Lu LF
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2017
TGF-β1 secreted by Tregs in lymph nodes promotes breast cancer malignancy via up-regulation of IL-17RB.
EMBO molecular medicine
Huang SC,Wei PC,Hwang-Verslues WW,Kuo WH,Jeng YM,Hu CM,Shew JY,Huang CS,Chang KJ,Lee EY,Lee WH
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Neutralization
Fri Dec 01 00:00:00 EST 2017
Development of effective tumor immunotherapy using a novel dendritic cell-targeting Toll-like receptor ligand.
PloS one
De Silva NH,Akazawa T,Wijewardana V,Inoue N,Oyamada M,Ohta A,Tachibana Y,Wijesekera DPH,Kuwamura M,Nishizawa Y,Itoh K,Izawa T,Hatoya S,Hasegawa T,Yamate J,Inaba T,Sugiura K
11-7101 was used in Flow cytometry/Cell sorting to show that the combined treatment gave rise to emphatic amelioration in DC-based cancer therapy.
Tue Dec 26 00:00:00 EST 2017
Maternal-Derived Hepatitis B Virus e Antigen Alters Macrophage Function in Offspring to Drive Viral Persistence after Vertical Transmission.
Immunity
Tian Y,Kuo CF,Akbari O,Ou JH
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Tue May 17 00:00:00 EDT 2016
Low-Dose Intestinal Trichuris muris Infection Alters the Lung Immune Microenvironment and Can Suppress Allergic Airway Inflammation.
Infection and immunity
Chenery AL,Antignano F,Burrows K,Scheer S,Perona-Wright G,Zaph C
11-7101 was used in Flow cytometry/Cell sorting to study crosstalk between the intestine and lung during disease, showing that low-dose intestinal Trichuris muris infection alters the lung immune microenvironment.
Mon Feb 01 00:00:00 EST 2016
Heterologous Immunity between Adenoviruses and Hepatitis C Virus: A New Paradigm in HCV Immunity and Vaccines.
PloS one
Singh S,Vedi S,Samrat SK,Li W,Kumar R,Agrawal B
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Tue Jul 19 00:00:00 EDT 2016
Heterologous Immunity between Adenoviruses and Hepatitis C Virus: A New Paradigm in HCV Immunity and Vaccines.
PloS one
Singh S,Vedi S,Samrat SK,Li W,Kumar R,Agrawal B
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Tue Jul 19 00:00:00 EDT 2016
NFATc1 supports imiquimod-induced skin inflammation by suppressing IL-10 synthesis in B cells.
Nature communications
Alrefai H,Muhammad K,Rudolf R,Pham DA,Klein-Hessling S,Patra AK,Avots A,Bukur V,Sahin U,Tenzer S,Goebeler M,Kerstan A,Serfling E
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Wed May 25 00:00:00 EDT 2016
Towards programming immune tolerance through geometric manipulation of phosphatidylserine.
Biomaterials
Roberts RA,Eitas TK,Byrne JD,Johnson BM,Short PJ,McKinnon KP,Reisdorf S,Luft JC,DeSimone JM,Ting JP
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2015
Tristetraprolin Limits Inflammatory Cytokine Production in Tumor-Associated Macrophages in an mRNA Decay-Independent Manner.
Cancer research
Kratochvill F,Gratz N,Qualls JE,Van De Velde LA,Chi H,Kovarik P,Murray PJ
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2015
Mouse bone marrow-derived mesenchymal stem cells inhibit leukemia/lymphoma cell proliferation in vitro and in a mouse model of allogeneic bone marrow transplant.
International journal of molecular medicine
Song N,Gao L,Qiu H,Huang C,Cheng H,Zhou H,Lv S,Chen L,Wang J
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2015
Mouse bone marrow-derived mesenchymal stem cells inhibit leukemia/lymphoma cell proliferation in vitro and in a mouse model of allogeneic bone marrow transplant.
International journal of molecular medicine
Song N,Gao L,Qiu H,Huang C,Cheng H,Zhou H,Lv S,Chen L,Wang J
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2015
Dimethyl sulfoxide inhibits spontaneous diabetes and autoimmune recurrence in non-obese diabetic mice by inducing differentiation of regulatory T cells.
Toxicology and applied pharmacology
Lin GJ,Sytwu HK,Yu JC,Chen YW,Kuo YL,Yu CC,Chang HM,Chan DC,Huang SH
11-7101 was used in Flow cytometry/Cell sorting to demonstrate that in vivo DMSO treatment suppresses spontaneous diabetes and autoimmune recurrence in NOD mice.
Thu Jan 15 00:00:00 EST 2015
Preemptive donor apoptotic cell infusions induce IFN-γ-producing myeloid-derived suppressor cells for cardiac allograft protection.
Journal of immunology (Baltimore, Md. : 1950)
Bryant J,Lerret NM,Wang JJ,Kang HK,Tasch J,Zhang Z,Luo X
11-7101 was used in Flow cytometry/Cell sorting to investigate the role of MDSCs in the graft protection provided by ECDI-SPs, showing that preemptive donor apoptotic cell infusions induce IFN-γ-producing MDSCs.
Sun Jun 15 00:00:00 EDT 2014
Nizatidine, a small molecular compound, enhances killed H5N1 vaccine cell-mediated responses and protects mice from lethal viral challenge.
Human vaccines & immunotherapeutics
Wang S,Wu B,Xue J,Wang M,Chen R,Wang B
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Mon Jan 05 00:00:00 EST 2015
BCG vaccine mediated reduction in the MHC-II expression of macrophages and dendritic cells is reversed by activation of Toll-like receptors 7 and 9.
Cellular immunology
Bakhru P,Sirisaengtaksin N,Soudani E,Mukherjee S,Khan A,Jagannath C
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2014
BCG vaccine mediated reduction in the MHC-II expression of macrophages and dendritic cells is reversed by activation of Toll-like receptors 7 and 9.
Cellular immunology
Bakhru P,Sirisaengtaksin N,Soudani E,Mukherjee S,Khan A,Jagannath C
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2014
Prevention or early cure of type 1 diabetes by intranasal administration of gliadin in NOD mice.
PloS one
Funda DP,Fundova P,Hansen AK,Buschard K
11-7101 was used in Flow cytometry/Cell sorting to investigate whether intranasal (i.n.) administration of gliadin or gluten may arrest the diabetogenic process, showing that i.n. administration of gliadin prevents type 1 diabetes in NOD mice.
Tue Jan 13 00:00:00 EST 2015
Genetic deletion of chemokine receptor Ccr7 exacerbates atherogenesis in ApoE-deficient mice.
Cardiovascular research
Wan W,Lionakis MS,Liu Q,Roffê E,Murphy PM
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
CX(3)CR1(+) macrophages support IL-22 production by innate lymphoid cells during infection with Citrobacter rodentium.
Mucosal immunology
Manta C,Heupel E,Radulovic K,Rossini V,Garbi N,Riedel CU,Niess JH
Published figure using IL-10 monoclonal antibody (Product # 11-7101-41) in Flow Cytometry
Tue Jan 01 00:00:00 EST 2013
Osteoclast activated FoxP3+ CD8+ T-cells suppress bone resorption in vitro.
PloS one
Buchwald ZS,Kiesel JR,DiPaolo R,Pagadala MS,Aurora R
11-7101 was used in Flow cytometry/Cell sorting to demonstrate the effects of activated CD8 T cell-produced cytokines on osteoclast activity.
Thu Nov 08 00:00:00 EST 2012
IL-10 regulates viral lung immunopathology during acute respiratory syncytial virus infection in mice.
PloS one
Loebbermann J,Schnoeller C,Thornton H,Durant L,Sweeney NP,Schuijs M,O'Garra A,Johansson C,Openshaw PJ
11-7101 was used in Flow cytometry/Cell sorting to study experimental respiratory syncytial virus infection in IL-10 deficient mice and mice treated with anti-IL-10 receptor antibody.
Mon Jul 23 00:00:00 EDT 2012
Infection with a helminth parasite attenuates autoimmunity through TGF-beta-mediated suppression of Th17 and Th1 responses.
Journal of immunology (Baltimore, Md. : 1950)
Walsh KP,Brady MT,Finlay CM,Boon L,Mills KH
11-7101 was used in Flow cytometry/Cell sorting to provide evidence that parasites can exert bystander immunosuppression of pathogenic T cells that mediate autoimmune diseases.
Sat Aug 01 00:00:00 EDT 2009
Ly49 C/I-dependent NKT cell-derived IL-10 is required for corneal graft survival and peripheral tolerance.
Journal of leukocyte biology
Watte CM,Nakamura T,Lau CH,Ortaldo JR,Stein-Streilein J
11-7101 was used in Flow cytometry/Cell sorting to test if the NK inhibitory receptor functionally contributed to tolerance development.
Tue Apr 01 00:00:00 EDT 2008
Deletion of both ICAM-1 and C3 enhances severity of experimental autoimmune encephalomyelitis compared to C3-deficient mice.
Neuroscience letters
Smith SS,Ludwig M,Wohler JE,Bullard DC,Szalai AJ,Barnum SR
11-7101 was used in Flow cytometry/Cell sorting to examine experimental autoimmune encephalitis in ICAM-1 x C3 double knockout mice.
Fri Sep 12 00:00:00 EDT 2008