Product References
Group 2 innate lymphoid cells suppress neuroinflammation and brain injury following intracerebral hemorrhage.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Liu M,Wang D,Xu L,Pan Y,Huang H,Li M,Liu Q
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2024
TL1A is an epithelial alarmin that cooperates with IL-33 for initiation of allergic airway inflammation.
The Journal of experimental medicine
Schmitt P,Duval A,Camus M,Lefrançais E,Roga S,Dedieu C,Ortega N,Bellard E,Mirey E,Mouton-Barbosa E,Burlet-Schiltz O,Gonzalez-de-Peredo A,Cayrol C,Girard JP
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Mon Jun 03 00:00:00 UTC 2024
Self-amplifying mRNA seasonal influenza vaccines elicit mouse neutralizing antibody and cell-mediated immunity and protect ferrets.
NPJ vaccines
Cheung M,Chang C,Rathnasinghe R,Rossignol E,Zhang Y,Ferrari A,Patel H,Huang Y,Sanchez Guillen M,Scalzo T,Lee C,Otten GR,Settembre EC,Music N,Palladino G,Wen Y
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Wed Oct 04 00:00:00 UTC 2023
Non-glycosylated G protein with CpG ODN provides robust protection against respiratory syncytial virus without inducing eosinophilia.
Frontiers in immunology
Kawahara E,Shibata T,Hirai T,Yoshioka Y
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Fri Jan 05 00:00:00 UTC 2024
Solar ultraviolet B radiation promotes α-MSH secretion to attenuate the function of ILC2s via the pituitary-lung axis.
Nature communications
Huang Y,Zhu L,Cheng S,Dai R,Huang C,Song Y,Peng B,Li X,Wen J,Gong Y,Hu Y,Qian L,Zhu L,Zhang F,Yu L,Yi C,Gu W,Ling Z,Ma L,Tang W,Peng L,Shi G,Zhang Y,Sun B
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Tue Sep 12 00:00:00 UTC 2023
Monitoring Group 2 Innate Lymphoid Cell Biology in Models of Lung Inflammation.
Bio-protocol
Badrani JH,Strohm AN,Haung YA,Doherty TA
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Thu Jul 20 00:00:00 UTC 2023
Fibroblastic reticular cells in lymph node potentiate white adipose tissue beiging through neuro-immune crosstalk in male mice.
Nature communications
Cheong LY,Wang B,Wang Q,Jin L,Kwok KHM,Wu X,Shu L,Lin H,Chung SK,Cheng KKY,Hoo RLC,Xu A
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Fri Mar 03 00:00:00 UTC 2023
Calcaratarin D, a labdane diterpenoid, attenuates mouse asthma via modulating alveolar macrophage function.
British journal of pharmacology
Liao W,Foo HYC,Tran TNQ,Chai CLL,Wong WSF
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Sat Apr 01 00:00:00 UTC 2023
Dermis resident macrophages orchestrate localized ILC2 eosinophil circuitries to promote non-healing cutaneous leishmaniasis.
Nature communications
Lee SH,Kang B,Kamenyeva O,Ferreira TR,Cho K,Khillan JS,Kabat J,Kelsall BL,Sacks DL
12-7133-41 was used in Flow Cytometry to demonstrate that dermis-resident macrophages are self-maintained as a replicative niche for L. major by orchestrating localized type 2 circuitries with type 2 innate lymphoid cells and eosinophils.
Wed Nov 29 00:00:00 UTC 2023
Heritable vaginal bacteria influence immune tolerance and relate to early-life markers of allergic sensitization in infancy.
Cell reports. Medicine
McCauley KE,Rackaityte E,LaMere B,Fadrosh DW,Fujimura KE,Panzer AR,Lin DL,Lynch KV,Halkias J,Mendoza VF,Burt TD,Bendixsen C,Barnes K,Kim H,Jones K,Ownby DR,Johnson CC,Seroogy CM,Gern JE,Boushey HA,Lynch SV
12-7133-41 was used in Flow cytometry/Cell sorting to investigate if bacterial and atopy heritability are intimately linked.
Tue Aug 16 00:00:00 UTC 2022
Mitochondrial transcription factor A in RORγt+ lymphocytes regulate small intestine homeostasis and metabolism.
Nature communications
Fu Z,Dean JW,Xiong L,Dougherty MW,Oliff KN,Chen ZE,Jobin C,Garrett TJ,Zhou L
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Thu Jul 22 00:00:00 UTC 2021
Synergistic effect of non-neutralizing antibodies and interferon-γ for cross-protection against influenza.
iScience
Shibuya M,Tamiya S,Kawai A,Hirai T,Cragg MS,Yoshioka Y
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Fri Oct 22 00:00:00 UTC 2021
Type 1 Cytotoxic T Cells Increase in Placenta after Intrauterine Inflammation.
Frontiers in immunology
Liu J,Liu Y,Panda S,Liu A,Lei J,Burd I
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Fri Dec 17 00:00:00 UTC 2021
Gut Microbial Metabolite Pravastatin Attenuates Intestinal Ischemia/Reperfusion Injury Through Promoting IL-13 Release From Type II Innate Lymphoid Cells via IL-33/ST2 Signaling.
Frontiers in immunology
Deng F,Hu JJ,Yang X,Sun QS,Lin ZB,Zhao BC,Yao ZW,Luo SD,Chen ZL,Liu Y,Yan ZZ,Li C,Liu WF,Liu KX
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Tue Dec 21 00:00:00 UTC 2021
6-Phosphogluconate dehydrogenase (6PGD), a key checkpoint in reprogramming of regulatory T cells metabolism and function.
eLife
Daneshmandi S,Cassel T,Higashi RM,Fan TW,Seth P
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Thu Oct 28 00:00:00 UTC 2021
ILC3s control airway inflammation by limiting T cell responses to allergens and microbes.
Cell reports
Teng F,Tachó-Piñot R,Sung B,Farber DL,Worgall S,Hammad H,Lambrecht BN,Hepworth MR,Sonnenberg GF
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Tue Nov 23 00:00:00 UTC 2021
Dual vaccination against IL-4 and IL-13 protects against chronic allergic asthma in mice.
Nature communications
Conde E,Bertrand R,Balbino B,Bonnefoy J,Stackowicz J,Caillot N,Colaone F,Hamdi S,Houmadi R,Loste A,Kamphuis JBJ,Huetz F,Guilleminault L,Gaudenzio N,Mougel A,Hardy D,Snouwaert JN,Koller BH,Serra V,Bruhns P,Grouard-Vogel G,Reber LL
12-7133-41 was used in Flow Cytometry to imply that dual IL-4/IL-13 vaccination may represent a cost-effective, long-term therapeutic strategy for the treatment of allergic asthma as demonstrated in mouse models, although additional studies are warranted to assess its safety and feasibility.
Tue May 11 00:00:00 UTC 2021
GM-CSF production by non-classical monocytes controls antagonistic LPS-driven functions in allergic inflammation.
Cell reports
Kaur K,Bachus H,Lewis C,Papillion AM,Rosenberg AF,Ballesteros-Tato A,León B
12-7133-41 was used in Flow cytometry/Cell sorting to show here that lipopolysaccharide activity switches from pro-pathogenic to protective depending on the production of granulocyte-macrophage colony-stimulating factor by non-classical monocytes.
Tue Dec 28 00:00:00 UTC 2021
Febrile Temperature Critically Controls the Differentiation and Pathogenicity of T Helper 17 Cells.
Immunity
Wang X,Ni L,Wan S,Zhao X,Ding X,Dejean A,Dong C
12-7133 was used in Flow cytometry/Cell sorting to show that febrile temperature selectively regulated Th17 cell differentiation in vitro in enhancing interleukin-17 (IL-17), IL-17F, and IL-22 expression.
Tue Feb 18 00:00:00 UTC 2020
A regulatory T cell Notch4-GDF15 axis licenses tissue inflammation in asthma.
Nature immunology
Harb H,Stephen-Victor E,Crestani E,Benamar M,Massoud A,Cui Y,Charbonnier LM,Arbag S,Baris S,Cunnigham A,Leyva-Castillo JM,Geha RS,Mousavi AJ,Guennewig B,Schmitz-Abe K,Sioutas C,Phipatanakul W,Chatila TA
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Sun Nov 01 00:00:00 UTC 2020
Bcl6 and Blimp1 reciprocally regulate ST2+ Treg-cell development in the context of allergic airway inflammation.
The Journal of allergy and clinical immunology
Koh B,Ulrich BJ,Nelson AS,Panangipalli G,Kharwadkar R,Wu W,Xie MM,Fu Y,Turner MJ,Paczesny S,Janga SC,Dent AL,Kaplan MH
12-7133 was used in Flow cytometry/Cell sorting to define the functions of Bcl6 in Treg cells, including Tfr cells, in the context of allergic airway inflammation.
Sun Nov 01 00:00:00 UTC 2020
Novel, Non-Gene-Destructive Knock-In Reporter Mice Refute the Concept of Monoallelic Gata3 Expression.
Journal of immunology (Baltimore, Md. : 1950)
Rao TN,Kumar S,Pulikkottil AJ,Oliveri F,Hendriks RW,Beckel F,Fehling HJ
12-7133 was used in Flow cytometry/Cell sorting to show that GATIR mice faithfully reflect endogenous Gata3 expression without disturbing the development of GATA-3-dependent lymphoid cell populations.
Fri May 01 00:00:00 UTC 2020
A helminth-derived suppressor of ST2 blocks allergic responses.
eLife
Vacca F,Chauché C,Jamwal A,Hinchy EC,Heieis G,Webster H,Ogunkanbi A,Sekne Z,Gregory WF,Wear M,Perona-Wright G,Higgins MK,Nys JA,Cohen ES,McSorley HJ
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Mon May 18 00:00:00 UTC 2020
Arf1-mediated lipid metabolism sustains cancer cells and its ablation induces anti-tumor immune responses in mice.
Nature communications
Wang G,Xu J,Zhao J,Yin W,Liu D,Chen W,Hou SX
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Fri Jan 10 00:00:00 UTC 2020
Polychromic Reporter Mice Reveal Unappreciated Innate Lymphoid Cell Progenitor Heterogeneity and Elusive ILC3 Progenitors in Bone Marrow.
Immunity
Walker JA,Clark PA,Crisp A,Barlow JL,Szeto A,Ferreira ACF,Rana BMJ,Jolin HE,Rodriguez-Rodriguez N,Sivasubramaniam M,Pannell R,Cruickshank J,Daly M,Haim-Vilmovsky L,Teichmann SA,McKenzie ANJ
12-7133 was used in Flow cytometry/Cell sorting to identify a discrete ILC2-committed population and delineated transition states between early progenitors and a highly heterogeneous ILC1, ILC3, and NK precursor cell cluster.
Tue Jul 16 00:00:00 UTC 2019
A Mutation in the Transcription Factor Foxp3 Drives T Helper 2 Effector Function in Regulatory T Cells.
Immunity
Van Gool F,Nguyen MLT,Mumbach MR,Satpathy AT,Rosenthal WL,Giacometti S,Le DT,Liu W,Brusko TM,Anderson MS,Rudensky AY,Marson A,Chang HY,Bluestone JA
12-7133 was used in Flow cytometry/Cell sorting to identify an IPEX (immune dysregulation polyendocrinopathy enteropathy X-linked) syndrome patient with a FOXP3 mutation in the domain swap interface of the protein.
Tue Feb 19 00:00:00 UTC 2019
Calcitonin Gene-Related Peptide Negatively Regulates Alarmin-Driven Type 2 Innate Lymphoid Cell Responses.
Immunity
Wallrapp A,Burkett PR,Riesenfeld SJ,Kim SJ,Christian E,Abdulnour RE,Thakore PI,Schnell A,Lambden C,Herbst RH,Khan P,Tsujikawa K,Xavier RJ,Chiu IM,Levy BD,Regev A,Kuchroo VK
12-7133-41 was used in Flow Cytometry to show that ILC2s expressed both the neuropeptide calcitonin gene-related peptide and its receptor.
Tue Oct 15 00:00:00 UTC 2019
IL-33 Induces Murine Intestinal Goblet Cell Differentiation Indirectly via Innate Lymphoid Cell IL-13 Secretion.
Journal of immunology (Baltimore, Md. : 1950)
Waddell A,Vallance JE,Hummel A,Alenghat T,Rosen MJ
12-7133 was used in Flow cytometry/Cell sorting to report that IL-33 does not directly induce goblet cell differentiation in murine enteroids; however, IL-13, a cytokine induced by IL-33, markedly induces goblet cells and gene expression consistent with goblet cell differentiation.
Tue Jan 15 00:00:00 UTC 2019
Ovalbumin induces natural killer cells to secrete Th2 cytokines IL‑5 and IL‑13 in a mouse model of asthma.
Molecular medicine reports
Chen Z,Wang L
12-7133-41 was used in Flow Cytometry to analyse the dynamic changes of NK cells and their subsets during the development of the ovalbumin (OVA)‚induced allergic airway response.
Mon Apr 01 00:00:00 UTC 2019
Costimulation of type-2 innate lymphoid cells by GITR promotes effector function and ameliorates type 2 diabetes.
Nature communications
Galle-Treger L,Sankaranarayanan I,Hurrell BP,Howard E,Lo R,Maazi H,Lewis G,Banie H,Epstein AL,Hu P,Rehan VK,Gilliland FD,Allayee H,Soroosh P,Sharpe AH,Akbari O
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Tue Feb 12 00:00:00 UTC 2019
TNFR2 Signaling Enhances ILC2 Survival, Function, and Induction of Airway Hyperreactivity.
Cell reports
Hurrell BP,Galle-Treger L,Jahani PS,Howard E,Helou DG,Banie H,Soroosh P,Akbari O
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Tue Dec 24 00:00:00 UTC 2019
Double negative T cells mediate Lag3-dependent antigen-specific protection in allergic asthma.
Nature communications
Tian D,Yang L,Wang S,Zhu Y,Shi W,Zhang C,Jin H,Tian Y,Xu H,Sun G,Liu K,Zhang Z,Zhang D
12-7133-41 was used in Flow cytometry/Cell sorting to investigate the therapeutic potential of adoptively transferring ovalbumin (OVA) peptide-primed CD4-CD8- double negative T (DNT) cells in a mouse model of OVA-induced allergic asthma.
Wed Sep 18 00:00:00 UTC 2019
The droplet size of emulsion adjuvants has significant impact on their potency, due to differences in immune cell-recruitment and -activation.
Scientific reports
Shah RR,Taccone M,Monaci E,Brito LA,Bonci A,O'Hagan DT,Amiji MM,Seubert A
12-7133 was used in Flow cytometry/Cell sorting to deduce the considerable impact of emulsion droplet size and composition on the ability to recruit and activate immune cells.
Thu Aug 08 00:00:00 UTC 2019
A chemical biology toolbox to study protein methyltransferases and epigenetic signaling.
Nature communications
Scheer S,Ackloo S,Medina TS,Schapira M,Li F,Ward JA,Lewis AM,Northrop JP,Richardson PL,Kaniskan HÜ,Shen Y,Liu J,Smil D,McLeod D,Zepeda-Velazquez CA,Luo M,Jin J,Barsyte-Lovejoy D,Huber KVM,De Carvalho DD,Vedadi M,Zaph C,Brown PJ,Arrowsmith CH
12-7133 was used in Flow cytometry/Cell sorting to present a collection of chemical probes and associated reagents and data to elucidate the function of human and murine protein methyltransferases in cellular studies.
Thu Jan 03 00:00:00 UTC 2019
Ets1 suppresses atopic dermatitis by suppressing pathogenic T cell responses.
JCI insight
Lee CG,Kwon HK,Kang H,Kim Y,Nam JH,Won YH,Park S,Kim T,Kang K,Rudra D,Jun CD,Park ZY,Im SH
12-7133 was used in Flow cytometry/Cell sorting to reveal a protective role of Ets1 in restricting pathogenic Th cell responses and suggest a potential therapeutic target for atopic dermatitis (AD) treatment.
Thu Mar 07 00:00:00 UTC 2019
Lung Single-Cell Signaling Interaction Map Reveals Basophil Role in Macrophage Imprinting.
Cell
Cohen M,Giladi A,Gorki AD,Solodkin DG,Zada M,Hladik A,Miklosi A,Salame TM,Halpern KB,David E,Itzkovitz S,Harkany T,Knapp S,Amit I
12-7133-41 was used in Flow Cytometry to demonstrate how whole-tissue signaling interaction map on the single-cell level can broaden our understanding of cellular networks in health and disease.
Thu Nov 01 00:00:00 UTC 2018
E3 Ligase VHL Promotes Group 2 Innate Lymphoid Cell Maturation and Function via Glycolysis Inhibition and Induction of Interleukin-33 Receptor.
Immunity
Li Q,Li D,Zhang X,Wan Q,Zhang W,Zheng M,Zou L,Elly C,Lee JH,Liu YC
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Tue Feb 20 00:00:00 UTC 2018
Autoimmune Th17 Cells Induced Synovial Stromal and Innate Lymphoid Cell Secretion of the Cytokine GM-CSF to Initiate and Augment Autoimmune Arthritis.
Immunity
Hirota K,Hashimoto M,Ito Y,Matsuura M,Ito H,Tanaka M,Watanabe H,Kondoh G,Tanaka A,Yasuda K,Kopf M,Potocnik AJ,Stockinger B,Sakaguchi N,Sakaguchi S
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Tue Jun 19 00:00:00 UTC 2018
Fli1-haploinsufficient dermal fibroblasts promote skin-localized transdifferentiation of Th2-like regulatory T cells.
Arthritis research & therapy
Saigusa R,Asano Y,Taniguchi T,Hirabayashi M,Nakamura K,Miura S,Yamashita T,Takahashi T,Ichimura Y,Toyama T,Yoshizaki A,Trojanowska M,Sato S
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Wed Feb 07 00:00:00 UTC 2018
Nematode-Infected Mice Acquire Resistance to Subsequent Infection With Unrelated Nematode by Inducing Highly Responsive Group 2 Innate Lymphoid Cells in the Lung.
Frontiers in immunology
Yasuda K,Adachi T,Koida A,Nakanishi K
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Mon Sep 30 00:00:00 UTC 2019
Genome-wide analyses reveal the IRE1a-XBP1 pathway promotes T helper cell differentiation by resolving secretory stress and accelerating proliferation.
Genome medicine
Pramanik J,Chen X,Kar G,Henriksson J,Gomes T,Park JE,Natarajan K,Meyer KB,Miao Z,McKenzie ANJ,Mahata B,Teichmann SA
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Wed Oct 24 00:00:00 UTC 2018
Fas Promotes T Helper 17 Cell Differentiation and Inhibits T Helper 1 Cell Development by Binding and Sequestering Transcription Factor STAT1.
Immunity
Meyer Zu Horste G,Przybylski D,Schramm MA,Wang C,Schnell A,Lee Y,Sobel R,Regev A,Kuchroo VK
12-7133-41 was used in Flow Cytometry to demonstrate that Fas promoted the generation and stability of Th17 cells and prevented their differentiation into Th1 cells.
Tue Mar 20 00:00:00 UTC 2018
mTOR complexes differentially orchestrates eosinophil development in allergy.
Scientific reports
Zhu C,Xia L,Li F,Zhou L,Weng Q,Li Z,Wu Y,Mao Y,Zhang C,Wu Y,Li M,Ying S,Chen Z,Shen H,Li W
12-7133 was used in Flow cytometry/Cell sorting to investigate the role of mTOR in eosinophil differentiation in the context of asthmatic pathogenesis.
Wed May 02 00:00:00 UTC 2018
Epidermal Growth Factor Receptor Expression Licenses Type-2 Helper T Cells to Function in a T Cell Receptor-Independent Fashion.
Immunity
Minutti CM,Drube S,Blair N,Schwartz C,McCrae JC,McKenzie AN,Kamradt T,Mokry M,Coffer PJ,Sibilia M,Sijts AJ,Fallon PG,Maizels RM,Zaiss DM
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Tue Oct 17 00:00:00 UTC 2017
HpARI Protein Secreted by a Helminth Parasite Suppresses Interleukin-33.
Immunity
Osbourn M,Soares DC,Vacca F,Cohen ES,Scott IC,Gregory WF,Smyth DJ,Toivakka M,Kemter AM,le Bihan T,Wear M,Hoving D,Filbey KJ,Hewitson JP,Henderson H,Gonzàlez-Cìscar A,Errington C,Vermeren S,Astier AL,Wallace WA,Schwarze J,Ivens AC,Maizels RM,McSorley HJ
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Tue Oct 17 00:00:00 UTC 2017
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Tue Oct 17 00:00:00 UTC 2017
Oxygen Sensing by T Cells Establishes an Immunologically Tolerant Metastatic Niche.
Cell
Clever D,Roychoudhuri R,Constantinides MG,Askenase MH,Sukumar M,Klebanoff CA,Eil RL,Hickman HD,Yu Z,Pan JH,Palmer DC,Phan AT,Goulding J,Gattinoni L,Goldrath AW,Belkaid Y,Restifo NP
12-7133 was used in Flow cytometry/Cell sorting to find that T-cell-intrinsic expression of the oxygen-sensing prolyl-hydroxylase proteins is required to maintain local tolerance against innocuous antigens in the lung but powerfully licenses colonization by circulating tumor cells.
Thu Aug 25 00:00:00 UTC 2016
Lack of autophagy induces steroid-resistant airway inflammation.
The Journal of allergy and clinical immunology
Suzuki Y,Maazi H,Sankaranarayanan I,Lam J,Khoo B,Soroosh P,Barbers RG,James Ou JH,Jung JU,Akbari O
12-7133 was used in Flow cytometry/Cell sorting to examine the role of autophagy in lung inflammation and the pathogenesis of corticosteroid-resistant neutrophilic asthma.
Sun May 01 00:00:00 UTC 2016
Transcription Factors Oct-1 and GATA-3 Cooperatively Regulate Th2 Cytokine Gene Expression via the RHS5 within the Th2 Locus Control Region.
PloS one
Kim K,Kim N,Lee GR
12-7133 was used in Flow cytometry/Cell sorting to investigate the molecular mechanisms behind Th2 cytokine gene regulation.
Thu Jul 07 00:00:00 UTC 2016
1,25D3 prevents CD8(+)Tc2 skewing and asthma development through VDR binding changes to the Cyp11a1 promoter.
Nature communications
Schedel M,Jia Y,Michel S,Takeda K,Domenico J,Joetham A,Ning F,Strand M,Han J,Wang M,Lucas JJ,Vogelberg C,Kabesch M,O'Connor BP,Gelfand EW
12-7133 was used in Flow cytometry/Cell sorting to investigate the underlying regulatory mechanisms of the conversion of Tc1 to Tc2 effector T cells in the presence of IL-4.
Mon Jan 11 00:00:00 UTC 2016
Bach2-Batf interactions control Th2-type immune response by regulating the IL-4 amplification loop.
Nature communications
Kuwahara M,Ise W,Ochi M,Suzuki J,Kometani K,Maruyama S,Izumoto M,Matsumoto A,Takemori N,Takemori A,Shinoda K,Nakayama T,Ohara O,Yasukawa M,Sawasaki T,Kurosaki T,Yamashita M
12-7133 was used in Flow cytometry/Cell sorting to suggest that IL-4 and Batf form a positive feedback amplification loop to induce Th2 cell differentiation and the subsequent Th2-type immune response.
Thu Sep 01 00:00:00 UTC 2016
Group 2 innate lymphoid cells promote beiging of white adipose tissue and limit obesity.
Nature
Brestoff JR,Kim BS,Saenz SA,Stine RR,Monticelli LA,Sonnenberg GF,Thome JJ,Farber DL,Lutfy K,Seale P,Artis D
12-7133 was used in Immunofluorescence to study how ILC2s can regulate adipose function and metabolic homeostasis.
Thu Mar 12 00:00:00 UTC 2015
Persistence of asthma requires multiple feedback circuits involving type 2 innate lymphoid cells and IL-33.
The Journal of allergy and clinical immunology
Christianson CA,Goplen NP,Zafar I,Irvin C,Good JT,Rollins DR,Gorentla B,Liu W,Gorska MM,Chu H,Martin RJ,Alam R
12-7133 was used in Flow cytometry/Cell sorting to present a critical network of feedback and feed-forward interactions between epithelial cells and ILC2s involved in maintaining chronic asthma.
Wed Jul 01 00:00:00 UTC 2015
Haploinsufficiency for Stard7 is associated with enhanced allergic responses in lung and skin.
Journal of immunology (Baltimore, Md. : 1950)
Yang L,Lewkowich I,Apsley K,Fritz JM,Wills-Karp M,Weaver TE
12-7133 was used in Flow cytometry/Cell sorting to determine whether loss of Stard7 expression promotes allergic responses, showing that haploinsufficiency for Stard7 is associated with enhanced allergic responses.
Mon Jun 15 00:00:00 UTC 2015
Triclosan Induces Thymic Stromal Lymphopoietin in Skin Promoting Th2 Allergic Responses.
Toxicological sciences : an official journal of the Society of Toxicology
Marshall NB,Lukomska E,Long CM,Kashon ML,Sharpnack DD,Nayak AP,Anderson KL,Jean Meade B,Anderson SE
12-7133 was used in Flow cytometry/Cell sorting to examine the effects of dermal triclosan exposure on the skin and lymph nodes of mice and in a human skin model to identify mechanisms for augmenting allergic responses.
Tue Sep 01 00:00:00 UTC 2015
The Effects of Antigen-Specific IgG1 Antibody for the Pulmonary-Hypertension-Phenotype and B Cells for Inflammation in Mice Exposed to Antigen and Fine Particles from Air Pollution.
PloS one
Park SH,Chen WC,Durmus N,Bleck B,Reibman J,Riemekasten G,Grunig G
12-7133 was used in Flow cytometry/Cell sorting to identify B cells and antigen specific IgG1 as potential therapeutical targets for pulmonary hypertension.
Mon Apr 25 00:00:00 UTC 2016
A microRNA upregulated in asthma airway T cells promotes TH2 cytokine production.
Nature immunology
Simpson LJ,Patel S,Bhakta NR,Choy DF,Brightbill HD,Ren X,Wang Y,Pua HH,Baumjohann D,Montoya MM,Panduro M,Remedios KA,Huang X,Fahy JV,Arron JR,Woodruff PG,Ansel KM
12-7133 was used in Flow cytometry/Cell sorting to study how upregulation of miR-19a in asthma may be an indicator and a cause of increased TH2 cytokine production in the airways.
Mon Dec 01 00:00:00 UTC 2014
Biliary repair and carcinogenesis are mediated by IL-33-dependent cholangiocyte proliferation.
The Journal of clinical investigation
Li J,Razumilava N,Gores GJ,Walters S,Mizuochi T,Mourya R,Bessho K,Wang YH,Glaser SS,Shivakumar P,Bezerra JA
12-7133 was used in Flow cytometry/Cell sorting to demonstrate the role of the Th2-promoting cytokine, IL-33, in cholangiocyte proliferation.
Tue Jul 01 00:00:00 UTC 2014
Natural killer cell NKG2D and granzyme B are critical for allergic pulmonary inflammation.
The Journal of allergy and clinical immunology
Farhadi N,Lambert L,Triulzi C,Openshaw PJ,Guerra N,Culley FJ
12-7133 was used in Flow cytometry/Cell sorting to investigate the importance of natural killer cells in house dust mite-triggered allergic response.
Sat Mar 01 00:00:00 UTC 2014
Immunosuppression by N-methyl-D-aspartate receptor antagonists is mediated through inhibition of Kv1.3 and KCa3.1 channels in T cells.
Molecular and cellular biology
Kahlfuß S,Simma N,Mankiewicz J,Bose T,Lowinus T,Klein-Hessling S,Sprengel R,Schraven B,Heine M,Bommhardt U
12-7133 was used in Flow cytometry/Cell sorting to investigate the effect of NMDAR antagonists on T-cell function, showing that antagonists cause immunosuppression via inhibition of Kv1.3 and KCa3.1 channels in T-cells.
Sat Mar 01 00:00:00 UTC 2014
Cryptococcus gattii infection dampens Th1 and Th17 responses by attenuating dendritic cell function and pulmonary chemokine expression in the immunocompetent hosts.
Infection and immunity
Angkasekwinai P,Sringkarin N,Supasorn O,Fungkrajai M,Wang YH,Chayakulkeeree M,Ngamskulrungroj P,Angkasekwinai N,Pattanapanyasat K
12-7133 was used in Flow cytometry/Cell sorting to investigate the impact of C. gattii infection on the development of the adaptive T helper cell immune response.
Mon Sep 01 00:00:00 UTC 2014
The Menin-Bach2 axis is critical for regulating CD4 T-cell senescence and cytokine homeostasis.
Nature communications
Kuwahara M,Suzuki J,Tofukuji S,Yamada T,Kanoh M,Matsumoto A,Maruyama S,Kometani K,Kurosaki T,Ohara O,Nakayama T,Yamashita M
12-7133 was used in Flow cytometry/Cell sorting to show that T cell-specific Menin deficiency results in the premature senescence of CD4 T cells, which is accompanied by the senescence-associated secretory phenotype after antigenic stimulation and dysregulated cytokine production.
Wed Apr 02 00:00:00 UTC 2014
Type 2 innate lymphoid cells control eosinophil homeostasis.
Nature
Nussbaum JC,Van Dyken SJ,von Moltke J,Cheng LE,Mohapatra A,Molofsky AB,Thornton EE,Krummel MF,Chawla A,Liang HE,Locksley RM
Published figure using IL-13 monoclonal antibody (Product # 12-7133-41) in Flow Cytometry
Thu Oct 10 00:00:00 UTC 2013
Interleukin-33-dependent innate lymphoid cells mediate hepatic fibrosis.
Immunity
McHedlidze T,Waldner M,Zopf S,Walker J,Rankin AL,Schuchmann M,Voehringer D,McKenzie AN,Neurath MF,Pflanz S,Wirtz S
12-7133 was used in Flow cytometry/Cell sorting to investigate the role of inflammatory networks in liver fibrosis, showing that it is mediated by interleukin-33-dependent innate lymphoid cells.
Thu Aug 22 00:00:00 UTC 2013
T cell activation induces proteasomal degradation of Argonaute and rapid remodeling of the microRNA repertoire.
The Journal of experimental medicine
Bronevetsky Y,Villarino AV,Eisley CJ,Barbeau R,Barczak AJ,Heinz GA,Kremmer E,Heissmeyer V,McManus MT,Erle DJ,Rao A,Ansel KM
12-7133 was used in Flow cytometry/Cell sorting to reveal T cell activation induces global post-transcriptional miRNA down-regulation, promoting acquisition of helper T cell effector functions.
Mon Feb 11 00:00:00 UTC 2013
Diesel exhaust particle induction of IL-17A contributes to severe asthma.
The Journal of allergy and clinical immunology
Brandt EB,Kovacic MB,Lee GB,Gibson AM,Acciani TH,Le Cras TD,Ryan PH,Budelsky AL,Khurana Hershey GK
12-7133 was used in Flow cytometry/Cell sorting to determine the mechanism by which diesel exhaust particles affect asthma severity, showing that diesel exhaust particles increase airway responsiveness to house dust mite extract due to increased levels of IL-17A, suggesting neutralisation of IL-17A could be used as a future therapy.
Fri Nov 01 00:00:00 UTC 2013
Steroidogenic enzyme Cyp11a1 regulates Type 2 CD8+ T cell skewing in allergic lung disease.
Proceedings of the National Academy of Sciences of the United States of America
Jia Y,Domenico J,Takeda K,Han J,Wang M,Armstrong M,Reisdorph N,O'Connor BP,Lucas JJ,Gelfand EW
12-7133 was used in Flow cytometry/Cell sorting to demonstrate that Cyp11a1 controls the phenotypic conversion of CD8+ T-cells from IFN-γ to IL-13 production.
Tue May 14 00:00:00 UTC 2013
Regulatory T cells prevent Th2 immune responses and pulmonary eosinophilia during respiratory syncytial virus infection in mice.
Journal of virology
Durant LR,Makris S,Voorburg CM,Loebbermann J,Johansson C,Openshaw PJ
12-7133 was used in Flow cytometry/Cell sorting to provide evidence that dysregulation of normal immune responses to viral infection may contribute to severe respiratory viral disease.
Tue Oct 01 00:00:00 UTC 2013
Stepwise epigenetic and phenotypic alterations poise CD8+ T cells to mediate airway hyperresponsiveness and inflammation.
Journal of immunology (Baltimore, Md. : 1950)
Jia Y,Takeda K,Han J,Joetham A,Marcus RA,Lucas JJ,O'Connor BP,Gelfand EW
12-7133 was used in Flow cytometry/Cell sorting to show that an atopic, IL-4 rich environment predisposes CD8 T cells to allergen-mediated pathogenic conversion to IL-13 production.
Mon Apr 15 00:00:00 UTC 2013
Modification of hemodynamic and immune responses to exposure with a weak antigen by the expression of a hypomorphic BMPR2 gene.
PloS one
Park SH,Chen WC,Hoffman C,Marsh LM,West J,Grunig G
12-7133 was used in Flow cytometry/Cell sorting to test the idea that Th2 responses to a mild antigen, together with the expression of a hypomorphic BMPR2 gene, would trigger pulmonary hypertension.
Mon Jul 22 00:00:00 UTC 2013
Virus-like particle-induced protection against MRSA pneumonia is dependent on IL-13 and enhancement of phagocyte function.
The American journal of pathology
Rynda-Apple A,Dobrinen E,McAlpine M,Read A,Harmsen A,Richert LE,Calverley M,Pallister K,Voyich J,Wiley JA,Johnson B,Young M,Douglas T,Harmsen AG
12-7133 was used in Flow cytometry/Cell sorting to investigate whether altering the lung environment in anticipation of insults provides a novel therapy for emerging infectious diseases.
Sun Jul 01 00:00:00 UTC 2012
Donor B cells in transplants augment clonal expansion and survival of pathogenic CD4+ T cells that mediate autoimmune-like chronic graft-versus-host disease.
Journal of immunology (Baltimore, Md. : 1950)
Young JS,Wu T,Chen Y,Zhao D,Liu H,Yi T,Johnston H,Racine J,Li X,Wang A,Todorov I,Zeng D
12-7133 was used in Flow cytometry/Cell sorting to investigate the mechanisms whereby donor B cells augment chronic graft-versus-host disease (cGVHD) pathogenesis, showing that B cells augment clonal expansion and survival of pathogenic CD4+ T cells that mediate autoimmune-like cGVHD.
Sun Jul 01 00:00:00 UTC 2012
Innate lymphoid cells promote lung-tissue homeostasis after infection with influenza virus.
Nature immunology
Monticelli LA,Sonnenberg GF,Abt MC,Alenghat T,Ziegler CG,Doering TA,Angelosanto JM,Laidlaw BJ,Yang CY,Sathaliyawala T,Kubota M,Turner D,Diamond JM,Goldrath AW,Farber DL,Collman RG,Wherry EJ,Artis D
12-7133 was used in Flow cytometry/Cell sorting to demonstrate a critical role for lung ILCs in restoring airway epithelial integrity and tissue homeostasis after viral replication.
Tue Nov 01 00:00:00 UTC 2011
IL-17-producing invariant NKT cells in lymphoid organs are recent thymic emigrants identified by neuropilin-1 expression.
Blood
Milpied P,Massot B,Renand A,Diem S,Herbelin A,Leite-de-Moraes M,Rubio MT,Hermine O
12-7133 was used in Flow cytometry/Cell sorting to determine the function of recent thymic emigrant invariant natural killer (RTE TiNKT) cells in naive mice.
Thu Sep 15 00:00:00 UTC 2011
EpiChIP: gene-by-gene quantification of epigenetic modification levels.
Nucleic acids research
Hebenstreit D,Gu M,Haider S,Turner DJ,Liò P,Teichmann SA
12-7133 was used in Flow cytometry/Cell sorting to develop a software package that analyses ChIP-seq data in order to quantify epigenetic modification levels for individual genes.
Tue Mar 01 00:00:00 UTC 2011
Requirement of GATA-binding protein 3 for II13 gene expression in IL-18-stimulated Th1 cells.
International immunology
Nakahira M,Nakanishi K
12-7133 was used in Flow cytometry/Cell sorting to elucidate the molecular mechanism by which gene expression pattern is altered in T(h)1 cells in response to IL-18, showing that Gata3 is required for this altered gene expression.
Thu Dec 01 00:00:00 UTC 2011
DX5+NKT cells display phenotypical and functional differences between spleen and liver as well as NK1.1-Balb/c and NK1.1+ C57Bl/6 mice.
BMC immunology
Werner JM,Busl E,Farkas SA,Schlitt HJ,Geissler EK,Hornung M
12-7133 was used in Immunocytochemistry to investigate the expression of several T cell and NK cell markers in spleen and liver DX5+NKT cell subsets in mice.
Fri Apr 29 00:00:00 UTC 2011
Costimulator B7-DC attenuates strong Th2 responses induced by Nippostrongylus brasiliensis.
Journal of immunology (Baltimore, Md. : 1950)
Ishiwata K,Watanabe N,Guo M,Tomihara K,Brumlik MJ,Yagita H,Pardoll D,Chen L,Shin T
12-7133 was used in Immunocytochemistry to demonstrate that the inhibition of Th2 responses by B7-DC occurs independently of programmed death 1 but conceivably acts through an as yet unknown alternative receptor that enhances Th1 responses.
Mon Feb 15 00:00:00 UTC 2010
Foxa2 programs Th2 cell-mediated innate immunity in the developing lung.
Journal of immunology (Baltimore, Md. : 1950)
Chen G,Wan H,Luo F,Zhang L,Xu Y,Lewkowich I,Wills-Karp M,Whitsett JA
12-7133 was used in Flow cytometry/Cell sorting to demonstrate that selective expression of Foxa2 plays a key role in regulating Th2 cell-mediated pulmonary inflammation.
Tue Jun 01 00:00:00 UTC 2010
Antigen aggregation decides the fate of the allergic immune response.
Journal of immunology (Baltimore, Md. : 1950)
Zaborsky N,Brunner M,Wallner M,Himly M,Karl T,Schwarzenbacher R,Ferreira F,Achatz G
12-7133 was used in Flow cytometry/Cell sorting to elucidate the molecular background of hypoallergenicity using Betula verrucosa.
Fri Jan 15 00:00:00 UTC 2010
Human CD8+CXCR3+ T cells have the same function as murine CD8+CD122+ Treg.
European journal of immunology
Shi Z,Okuno Y,Rifa'i M,Endharti AT,Akane K,Isobe K,Suzuki H
12-7133-41 was used in Flow Cytometry to suggest that human CD8(+)CXCR3(+) T cells are the counterparts of murine CD8(+)CD122(+) Treg.
Sat Aug 01 00:00:00 UTC 2009