Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: HepG2. The target is usually found in the following locations: Cytoplasm, Nucleus, Secreted.
Immunogen sequence: MESHSRAGKS RKSAKFRSIS RSLMLCNAKT SDDGSSPDEK YPDPFEISLA QGKEGIFHSS VQLADTSEAG PSSVPDLALA SEAAQLQAAG NDRGKTCRRI FFMKESSTAS SREKPGKLEA QSSNFLFPKA CHQRARSNST SVNPYCTREI DFPMTKKSAA PTDRQPYSLC SNRKSLSQQL DCPAGKAAGT SRPTRSLSTA QLVQPSGGLQ ASVISNIVLM KGQAKGLGFS IVGGKDSIYG PIGIYVKTIF AGGAAAADGR LQEGDEILEL NGESMAGLTH QDALQKFKQA KKGLLTLTVR TRLTA
The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; ILN; Interleukin; Interleukin16; lymphocyte chemoattractant factor; M-IL-16; neuronal interleukin 16; Pro-interleukin-16; prointerleukin 16
Gene Aliases: IL16; LCF; mKIAA4048; NIL16; prIL-16; PRIL16
UniProt ID: (Human) Q14005, (Mouse) O54824
Entrez Gene ID: (Human) 3603, (Mouse) 16170
Molecular Function:
interleukin superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support