Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
In direct ELISAs, approximately 40% reactivity with recombinant mouse (rm) IL-17A/IL-17F heterodimer is observed. No cross-reactivity with recombinant human IL-17, recombinant canine IL-17, rmIL-17B, rmIL-17C, rmIL-17D, rmIL-17E, or rmIL-17F is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.
IL-17A and IL-17F are well-characterized homodimeric cytokines secreted by T helper 17 (Th17) cells, gamma-delta T cells and several subsets of innate lymphoid cells. Somewhat less appreciated, IL-17A and IL-17F subunits can also form the heterodimer, IL-17AF. Together, these three dimers signal through the IL-17RA/IL-17RC receptor complex to mediate immune responses at mucosal interfaces and are found at lesion sites in inflammatory bowel disease, asthma, atopic dermatitis and rheumatoid arthritis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTLA-8; Cytotoxic T-lymphocyte-associated antigen 8; il 17; IL-17; IL-17F; ILN; Interleukin; Interleukin-17A; Interleukin-17F
Gene Aliases: C87042; Ctla-8; Ctla8; IL-17; IL-17A; IL-17F; Il17; Il17a; Il17f
UniProt ID: (Mouse) Q62386, (Mouse) Q7TNI7
Entrez Gene ID: (Mouse) 16171, (Mouse) 257630
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support