Product References
Cathepsin S inhibition in dendritic cells prevents Th17 cell differentiation in perivascular adipose tissues following vascular injury in diabetic rats.
Journal of biochemical and molecular toxicology
Peng H,Lv Y,Li C,Cheng Z,He S,Wang C,Liu J
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Fri Sep 01 00:00:00 EDT 2023
FABP4 facilitates inflammasome activation to induce the Treg/Th17 imbalance in preeclampsia via forming a positive feedback with IL-17A.
Molecular therapy. Nucleic acids
Chang GP,Yang XL,Liu W,Lin S,Yang SL,Zhao MY
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Fri Jun 04 00:00:00 EDT 2021
Small molecule allosteric inhibitors of RORγt block Th17-dependent inflammation and associated gene expression in vivo.
PloS one
Saenz SA,Local A,Carr T,Shakya A,Koul S,Hu H,Chourb L,Stedman J,Malley J,D'Agostino LA,Shanmugasundaram V,Malona J,Schwartz CE,Beebe L,Clements M,Rajaraman G,Cho J,Jiang L,Dubrovskiy A,Kreilein M,Shimanovich R,Hamann LG,Escoubet L,Ellis JM
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Wed Dec 29 00:00:00 EST 2021
YY1 regulation by miR-124-3p promotes Th17 cell pathogenicity through interaction with T-bet in rheumatoid arthritis.
JCI insight
Lin J,Tang J,Lin J,He Y,Yu Z,Jiang R,Yang B,Ou Q
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Mon Nov 22 00:00:00 EST 2021
Neutrophil-Derived IL-17 Promotes Ventilator-Induced Lung Injury via p38 MAPK/MCP-1 Pathway Activation.
Frontiers in immunology
Liao X,Zhang W,Dai H,Jing R,Ye M,Ge W,Pei S,Pan L
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Immunohistochemistry (Paraffin)
Mon Feb 14 00:00:00 EST 2022
Neutrophil-Derived IL-17 Promotes Ventilator-Induced Lung Injury via p38 MAPK/MCP-1 Pathway Activation.
Frontiers in immunology
Liao X,Zhang W,Dai H,Jing R,Ye M,Ge W,Pei S,Pan L
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Immunohistochemistry (Paraffin)
Mon Feb 14 00:00:00 EST 2022
Th17/Treg imbalance modulates rat myocardial fibrosis and heart failure by regulating LOX expression.
Acta physiologica (Oxford, England)
Lu M,Qin X,Yao J,Yang Y,Zhao M,Sun L
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Neutralization
Sun Nov 01 00:00:00 EDT 2020
Activation of Human Vδ2+ γδ T Cells by Staphylococcus aureus Promotes Enhanced Anti-Staphylococcal Adaptive Immunity.
Journal of immunology (Baltimore, Md. : 1950)
Cooper AJR,Lalor SJ,McLoughlin RM
14-7179-82 was used in Flow Cytometry to study the capacity of human Vδ2+ γδ T cells for rapid activation in response to S. aureus.
Sat Aug 15 00:00:00 EDT 2020
Senolytics prevent mt-DNA-induced inflammation and promote the survival of aged organs following transplantation.
Nature communications
Iske J,Seyda M,Heinbokel T,Maenosono R,Minami K,Nian Y,Quante M,Falk CS,Azuma H,Martin F,Passos JF,Niemann CU,Tchkonia T,Kirkland JL,Elkhal A,Tullius SG
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Thu Aug 27 00:00:00 EDT 2020
Developing Human Skin Contains Lymphocytes Demonstrating a Memory Signature.
Cell reports. Medicine
Dhariwala MO,Karthikeyan D,Vasquez KS,Farhat S,Weckel A,Taravati K,Leitner EG,Clancy S,Pauli M,Piper ML,Cohen JN,Ashouri JF,Lowe MM,Rosenblum MD,Scharschmidt TC
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Tue Nov 17 00:00:00 EST 2020
T cell-intrinsic role for Nod2 in protection against Th17-mediated uveitis.
Nature communications
Napier RJ,Lee EJ,Davey MP,Vance EE,Furtado JM,Snow PE,Samson KA,Lashley SJ,Brown BR,Horai R,Mattapallil MJ,Xu B,Callegan MC,Uebelhoer LS,Lancioni CL,Vehe RK,Binstadt BA,Smith JR,Caspi RR,Rosenzweig HL
14-7179-82 was used in Immunocytochemistry to reveal a non-conventional, T cell-intrinsic function for Nod2 in suppression of Th17 immunity and experimental uveitis.
Mon Oct 26 00:00:00 EDT 2020
CD161 contributes to prenatal immune suppression of IFNγ-producing PLZF+ T cells.
The Journal of clinical investigation
Halkias J,Rackaityte E,Hillman SL,Aran D,Mendoza VF,Marshall LR,MacKenzie TC,Burt TD
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Thu May 30 00:00:00 EDT 2019
Reciprocal regulation of Th2 and Th17 cells by PAD2-mediated citrullination.
JCI insight
Sun B,Chang HH,Salinger A,Tomita B,Bawadekar M,Holmes CL,Shelef MA,Weerapana E,Thompson PR,Ho IC
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in ELISA
Thu Nov 14 00:00:00 EST 2019
CXCL4 is a novel inducer of human Th17 cells and correlates with IL-17 and IL-22 in psoriatic arthritis.
European journal of immunology
Affandi AJ,Silva-Cardoso SC,Garcia S,Leijten EFA,van Kempen TS,Marut W,van Roon JAG,Radstake TRDJ
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Thu Mar 01 00:00:00 EST 2018
Accumulation of T-helper 22 cells, interleukin-22 and myeloid-derived suppressor cells promotes gastric cancer progression in elderly patients.
Oncology letters
Chen X,Wang Y,Wang J,Wen J,Jia X,Wang X,Zhang H
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2018
Cutting Edge: Selective Oral ROCK2 Inhibitor Reduces Clinical Scores in Patients with Psoriasis Vulgaris and Normalizes Skin Pathology via Concurrent Regulation of IL-17 and IL-10.
Journal of immunology (Baltimore, Md. : 1950)
Zanin-Zhorov A,Weiss JM,Trzeciak A,Chen W,Zhang J,Nyuydzefe MS,Arencibia C,Polimera S,Schueller O,Fuentes-Duculan J,Bonifacio KM,Kunjravia N,Cueto I,Soung J,Fleischmann RM,Kivitz A,Lebwohl M,Nunez M,Woodson J,Smith SL,West RF,Berger M,Krueger JG,Ryan JL,Waksal SD
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in ELISA
Mon May 15 00:00:00 EDT 2017
Enhancing the toolbox to study IL-17A in cattle and sheep.
Veterinary research
Wattegedera SR,Corripio-Miyar Y,Pang Y,Frew D,McNeilly TN,Palarea-Albaladejo J,McInnes CJ,Hope JC,Glass EJ,Entrican G
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Sat Apr 08 00:00:00 EDT 2017
Enhancing the toolbox to study IL-17A in cattle and sheep.
Veterinary research
Wattegedera SR,Corripio-Miyar Y,Pang Y,Frew D,McNeilly TN,Palarea-Albaladejo J,McInnes CJ,Hope JC,Glass EJ,Entrican G
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Sat Apr 08 00:00:00 EDT 2017
Berberine exerts an anti-inflammatory role in ocular Behcet's disease.
Molecular medicine reports
Yang Y,Wang Q,Xie M,Liu P,Qi X,Liu X,Li Z
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Sun Jan 01 00:00:00 EST 2017
Transcriptional signature of human pro-inflammatory TH17 cells identifies reduced IL10 gene expression in multiple sclerosis.
Nature communications
Hu D,Notarbartolo S,Croonenborghs T,Patel B,Cialic R,Yang TH,Aschenbrenner D,Andersson KM,Gattorno M,Pham M,Kivisakk P,Pierre IV,Lee Y,Kiani K,Bokarewa M,Tjon E,Pochet N,Sallusto F,Kuchroo VK,Weiner HL
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Fri Nov 17 00:00:00 EST 2017
Single-cell profiling reveals GPCR heterogeneity and functional patterning during neuroinflammation.
JCI insight
Tischner D,Grimm M,Kaur H,Staudenraus D,Carvalho J,Looso M,Günther S,Wanke F,Moos S,Siller N,Breuer J,Schwab N,Zipp F,Waisman A,Kurschus FC,Offermanns S,Wettschureck N
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Thu Aug 03 00:00:00 EDT 2017
Commensal-Specific CD4(+) Cells From Patients With Crohn's Disease Have a T-Helper 17 Inflammatory Profile.
Gastroenterology
Calderón-Gómez E,Bassolas-Molina H,Mora-Buch R,Dotti I,Planell N,Esteller M,Gallego M,Martí M,Garcia-Martín C,Martínez-Torró C,Ordás I,Singh S,Panés J,Benítez-Ribas D,Salas A
14-7179 was used in Flow cytometry/Cell sorting to investigate the antigen-specificity of T cells isolated from patients with Crohn's disease.
Thu Sep 01 00:00:00 EDT 2016
CD16+ Monocyte Subset Was Enriched and Functionally Exacerbated in Driving T-Cell Activation and B-Cell Response in Systemic Lupus Erythematosus.
Frontiers in immunology
Zhu H,Hu F,Sun X,Zhang X,Zhu L,Liu X,Li X,Xu L,Shi L,Gan Y,Su Y
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
CD16+ Monocyte Subset Was Enriched and Functionally Exacerbated in Driving T-Cell Activation and B-Cell Response in Systemic Lupus Erythematosus.
Frontiers in immunology
Zhu H,Hu F,Sun X,Zhang X,Zhu L,Liu X,Li X,Xu L,Shi L,Gan Y,Su Y
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
Cytomegalovirus Infection Leads to Development of High Frequencies of Cytotoxic Virus-Specific CD4+ T Cells Targeted to Vascular Endothelium.
PLoS pathogens
Pachnio A,Ciaurriz M,Begum J,Lal N,Zuo J,Beggs A,Moss P
14-7179 was used in Flow cytometry/Cell sorting to reveal the marked accumulation and unique phenotype of CMV-specific CD4+ T cells.
Thu Sep 01 00:00:00 EDT 2016
Chemokine Levels in the Penile Coronal Sulcus Correlate with HIV-1 Acquisition and Are Reduced by Male Circumcision in Rakai, Uganda.
PLoS pathogens
Prodger JL,Gray RH,Shannon B,Shahabi K,Kong X,Grabowski K,Kigozi G,Nalugoda F,Serwadda D,Wawer MJ,Reynolds SJ,Liu CM,Tobian AA,Kaul R
14-7179 was used in Flow cytometry/Cell sorting to suggest that penile inflammation can increase HIV susceptibility and can be reduced by circumcision.
Tue Nov 01 00:00:00 EDT 2016
Role of podoplanin in the high interleukin-17A secretion resulting from interactions between activated lymphocytes and psoriatic skin-derived mesenchymal cells.
Clinical and experimental immunology
Noack M,Ndongo-Thiam N,Miossec P
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Sat Oct 01 00:00:00 EDT 2016
Interaction among activated lymphocytes and mesenchymal cells through podoplanin is critical for a high IL-17 secretion.
Arthritis research & therapy
Noack M,Ndongo-Thiam N,Miossec P
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Thu Jun 23 00:00:00 EDT 2016
Reduced pro-inflammatory profile of γδT cells in pregnant patients with rheumatoid arthritis.
Arthritis research & therapy
Tham M,Schlör GR,Yerly D,Mueller C,Surbek D,Villiger PM,Förger F
14-7179 was used in Flow cytometry/Cell sorting to investigate whether pregnancy-related changes in disease activity were associated with changes in the proportion and function of γδT cells.
Fri Jan 22 00:00:00 EST 2016
Altered expression of miR-92a correlates with Th17 cell frequency in patients with primary biliary cirrhosis.
International journal of molecular medicine
Liang DY,Hou YQ,Luo LJ,Ao L
14-7179 was used in Immunohistochemistry to study the altered expression of miR-92a and its impact on the frequency of Th17 cells in patients with primary biliary cirrhosis.
Fri Jul 01 00:00:00 EDT 2016
Altered expression of miR-92a correlates with Th17 cell frequency in patients with primary biliary cirrhosis.
International journal of molecular medicine
Liang DY,Hou YQ,Luo LJ,Ao L
14-7179 was used in Immunohistochemistry to study the altered expression of miR-92a and its impact on the frequency of Th17 cells in patients with primary biliary cirrhosis.
Fri Jul 01 00:00:00 EDT 2016
Vaccination against canine leishmaniosis increases the phagocytic activity, nitric oxide production and expression of cell activation/migration molecules in neutrophils and monocytes.
Veterinary parasitology
Moreira ML,Costa-Pereira C,Alves ML,Marteleto BH,Ribeiro VM,Peruhype-Magalhães V,Giunchetti RC,Martins-Filho OA,Araújo MS
14717982 was used in flow cytometry to assess the protective immunological events induced by vaccination with Leishmune in dogs
Fri Apr 15 00:00:00 EDT 2016
Brief Report: IFIH1 Mutation Causes Systemic Lupus Erythematosus With Selective IgA Deficiency.
Arthritis & rheumatology (Hoboken, N.J.)
Van Eyck L,De Somer L,Pombal D,Bornschein S,Frans G,Humblet-Baron S,Moens L,de Zegher F,Bossuyt X,Wouters C,Liston A
14-7179 was used in Flow cytometry/Cell sorting to determine the underlying genetic defect responsible for severe early onset and refractory systemic lupus erythematosus.
Mon Jun 01 00:00:00 EDT 2015
IL-17A production in human psoriatic blood and lesions by CD146+ T cells.
The Journal of investigative dermatology
Mehta NN,Dagur PK,Rose SM,Naik HB,Stansky E,Doveikis J,Biancotto A,Playford MP,Philip McCoy J
14-7179 was used in Immunocytochemistry to study IL-17A production in human psoriatic blood and lesions by CD146+ T cells.
Thu Jan 01 00:00:00 EST 2015
Human CD8+ T-cells recognizing peptides from Mycobacterium tuberculosis (Mtb) presented by HLA-E have an unorthodox Th2-like, multifunctional, Mtb inhibitory phenotype and represent a novel human T-cell subset.
PLoS pathogens
van Meijgaarden KE,Haks MC,Caccamo N,Dieli F,Ottenhoff TH,Joosten SA
14-7179 was used in Flow cytometry/Cell sorting to identify a novel human T-cell subset with an unorthodox, multifunctional Th2 like phenotype, involved in the human immune response to mycobacteria.
Sun Mar 01 00:00:00 EST 2015
Interleukin-25 Mediates Transcriptional Control of PD-L1 via STAT3 in Multipotent Human Mesenchymal Stromal Cells (hMSCs) to Suppress Th17 Responses.
Stem cell reports
Wang WB,Yen ML,Liu KJ,Hsu PJ,Lin MH,Chen PM,Sudhir PR,Chen CH,Chen CH,Sytwu HK,Yen BL
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Tue Sep 08 00:00:00 EDT 2015
Fiber composite slices for multiplexed immunoassays.
Biomicrofluidics
Kim J,Bae S,Song S,Chung K,Kwon S
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Immunofluorescence
Wed Jul 01 00:00:00 EDT 2015
Fiber composite slices for multiplexed immunoassays.
Biomicrofluidics
Kim J,Bae S,Song S,Chung K,Kwon S
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Immunofluorescence
Wed Jul 01 00:00:00 EDT 2015
Fiber composite slices for multiplexed immunoassays.
Biomicrofluidics
Kim J,Bae S,Song S,Chung K,Kwon S
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Immunofluorescence
Wed Jul 01 00:00:00 EDT 2015
Antigen-Specific Mammary Inflammation Depends on the Production of IL-17A and IFN-γ by Bovine CD4+ T Lymphocytes.
PloS one
Rainard P,Cunha P,Ledresseur M,Staub C,Touzé JL,Kempf F,Gilbert FB,Foucras G
14-7179 was used in Flow cytometry/Cell sorting to indicate that the antigen-specific WBA, and specifically IL-17A and IFN-γ production by circulating CD4+ cells, can be used as a predictor of mammary hypersensitivity to protein antigens.
Thu May 26 00:00:00 EDT 2016
Autosomal dominant immune dysregulation syndrome in humans with CTLA4 mutations.
Nature medicine
Schubert D,Bode C,Kenefeck R,Hou TZ,Wing JB,Kennedy A,Bulashevska A,Petersen BS,Schäffer AA,Grüning BA,Unger S,Frede N,Baumann U,Witte T,Schmidt RE,Dueckers G,Niehues T,Seneviratne S,Kanariou M,Speckmann C,Ehl S,Rensing-Ehl A,Warnatz K,Rakhmanov M,Thimme R,Hasselblatt P,Emmerich F,Cathomen T,Backofen R,Fisch P,Seidl M,May A,Schmitt-Graeff A,Ikemizu S,Salzer U,Franke A,Sakaguchi S,Walker LSK,Sansom DM,Grimbacher B
14-7179 was used in Flow cytometry/Cell sorting to demonstrate the consequences for mice with mutations in CTLA4, an essential negative regulator of immune responses.
Mon Dec 01 00:00:00 EST 2014
Immune correlates of HIV exposure without infection in foreskins of men from Rakai, Uganda.
Mucosal immunology
Prodger JL,Hirbod T,Kigozi G,Nalugoda F,Reynolds SJ,Galiwango R,Shahabi K,Serwadda D,Wawer MJ,Gray RH,Kaul R
14-7179 was used in Flow cytometry/Cell sorting to show that HESN foreskins had elevated α-defensins and HIV-neutralizing IgAs.
Thu May 01 00:00:00 EDT 2014
Hemin controls T cell polarization in sickle cell alloimmunization.
Journal of immunology (Baltimore, Md. : 1950)
Zhong H,Bao W,Friedman D,Yazdanbakhsh K
14-7179 was used in Flow cytometry/Cell sorting to investigate whether monocyte control of T cell polarisation differs between alloimmunised and non-alloimmunised sickle cell disease patients.
Tue Jul 01 00:00:00 EDT 2014
Skin-resident T cells sense ultraviolet radiation-induced injury and contribute to DNA repair.
Journal of immunology (Baltimore, Md. : 1950)
MacLeod AS,Rudolph R,Corriden R,Ye I,Garijo O,Havran WL
14-7179 was used in Flow cytometry/Cell sorting to establish a novel role for skin-resident T cells in the UVR-associated DNA repair response and underscore the importance of skin-resident T cells to overall skin regeneration.
Sun Jun 15 00:00:00 EDT 2014
Akt-dependent enhanced migratory capacity of Th17 cells from children with lupus nephritis.
Journal of immunology (Baltimore, Md. : 1950)
Kshirsagar S,Riedl M,Billing H,Tönshoff B,Thangavadivel S,Steuber C,Staude H,Wechselberger G,Edelbauer M
14-7179 was used in Flow cytometry/Cell sorting to determine that therapeutic modulation of Akt activity could suppress chronic inflammatory processes in children with lupus.
Sat Nov 15 00:00:00 EST 2014
Potential role of IL-17-producing iNKT cells in type 1 diabetes.
PloS one
Li S,Joseph C,Becourt C,Klibi J,Luce S,Dubois-Laforgue D,Larger E,Boitard C,Benlagha K
14-7179 was used in Flow cytometry/Cell sorting to investigate the status and potential role of iNKT17 in type 1 diabetes.
Thu Jan 22 00:00:00 EST 2015
An ACT1 mutation selectively abolishes interleukin-17 responses in humans with chronic mucocutaneous candidiasis.
Immunity
Boisson B,Wang C,Pedergnana V,Wu L,Cypowyj S,Rybojad M,Belkadi A,Picard C,Abel L,Fieschi C,Puel A,Li X,Casanova JL
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Thu Oct 17 00:00:00 EDT 2013
CD40L expression permits CD8+ T cells to execute immunologic helper functions.
Blood
Frentsch M,Stark R,Matzmohr N,Meier S,Durlanik S,Schulz AR,Stervbo U,Jürchott K,Gebhardt F,Heine G,Reuter MA,Betts MR,Busch D,Thiel A
14-7179 was used in Flow cytometry/Cell sorting to investigate the role of CD40L in CD8+ T cells, showing that expression permits CD8+ T cells to execute immunologic helper functions.
Thu Jul 18 00:00:00 EDT 2013
Patients with cystic fibrosis have inducible IL-17+IL-22+ memory cells in lung draining lymph nodes.
The Journal of allergy and clinical immunology
Chan YR,Chen K,Duncan SR,Lathrop KL,Latoche JD,Logar AJ,Pociask DA,Wahlberg BJ,Ray P,Ray A,Pilewski JM,Kolls JK
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Immunofluorescence
Mon Apr 01 00:00:00 EDT 2013
Ovine skin-recirculating γδ T cells express IFN-γ and IL-17 and exit tissue independently of CCR7.
Veterinary immunology and immunopathology
Geherin SA,Lee MH,Wilson RP,Debes GF
14-7179 was used in Flow cytometry/Cell sorting to suggest that γδ T cells survey skin and sites of inflammation and infection, entering and leaving via independent pathways.
Sun Sep 01 00:00:00 EDT 2013
Increased frequencies of Th22 cells as well as Th17 cells in the peripheral blood of patients with ankylosing spondylitis and rheumatoid arthritis.
PloS one
Zhang L,Li YG,Li YH,Qi L,Liu XG,Yuan CZ,Hu NW,Ma DX,Li ZF,Yang Q,Li W,Li JM
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Fri Jul 27 00:00:00 EDT 2012
Renal IL-17 expression in human ANCA-associated glomerulonephritis.
American journal of physiology. Renal physiology
Velden J,Paust HJ,Hoxha E,Turner JE,Steinmetz OM,Wolf G,Jabs WJ,Özcan F,Beige J,Heering PJ,Schröder S,Kneißler U,Disteldorf E,Mittrücker HW,Stahl RA,Helmchen U,Panzer U
14-7179 was used in Immunofluorescence to analyse human kidney biopsies of active necrotising and crescentic ANCA-associated glomerulonephritis.
Fri Jun 15 00:00:00 EDT 2012
Chronic mucocutaneous candidiasis in humans with inborn errors of interleukin-17 immunity.
Science (New York, N.Y.)
Puel A,Cypowyj S,Bustamante J,Wright JF,Liu L,Lim HK,Migaud M,Israel L,Chrabieh M,Audry M,Gumbleton M,Toulon A,Bodemer C,El-Baghdadi J,Whitters M,Paradis T,Brooks J,Collins M,Wolfman NM,Al-Muhsen S,Galicchio M,Abel L,Picard C,Casanova JL
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in ELISA
Fri Apr 01 00:00:00 EDT 2011
Characterization of interleukin-17-producing regulatory T cells in inflamed intestinal mucosa from patients with inflammatory bowel diseases.
Gastroenterology
Hovhannisyan Z,Treatman J,Littman DR,Mayer L
14-7179 was used in Flow cytometry/Cell sorting to investigate how Th17 and Tregs interact under chronic inflammatory conditions.
Tue Mar 01 00:00:00 EST 2011
Gain-of-function human STAT1 mutations impair IL-17 immunity and underlie chronic mucocutaneous candidiasis.
The Journal of experimental medicine
Liu L,Okada S,Kong XF,Kreins AY,Cypowyj S,Abhyankar A,Toubiana J,Itan Y,Audry M,Nitschke P,Masson C,Toth B,Flatot J,Migaud M,Chrabieh M,Kochetkov T,Bolze A,Borghesi A,Toulon A,Hiller J,Eyerich S,Eyerich K,Gulácsy V,Chernyshova L,Chernyshov V,Bondarenko A,Grimaldo RM,Blancas-Galicia L,Beas IM,Roesler J,Magdorf K,Engelhard D,Thumerelle C,Burgel PR,Hoernes M,Drexel B,Seger R,Kusuma T,Jansson AF,Sawalle-Belohradsky J,Belohradsky B,Jouanguy E,Bustamante J,Bué M,Karin N,Wildbaum G,Bodemer C,Lortholary O,Fischer A,Blanche S,Al-Muhsen S,Reichenbach J,Kobayashi M,Rosales FE,Lozano CT,Kilic SS,Oleastro M,Etzioni A,Traidl-Hoffmann C,Renner ED,Abel L,Picard C,Maródi L,Boisson-Dupuis S,Puel A,Casanova JL
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2011
Central nervous system recruitment of effector memory CD8+ T lymphocytes during neuroinflammation is dependent on α4 integrin.
Brain : a journal of neurology
Ifergan I,Kebir H,Alvarez JI,Marceau G,Bernard M,Bourbonnière L,Poirier J,Duquette P,Talbot PJ,Arbour N,Prat A
14-7179 was used in Flow cytometry/Cell sorting to demonstrate a role of the blood-brain barrier in the recruitment of effector memory CD8(+) T lymphocytes to the CNS compartment.
Thu Dec 01 00:00:00 EST 2011
CD4-like immunological function by CD4- T cells in multiple natural hosts of simian immunodeficiency virus.
Journal of virology
Vinton C,Klatt NR,Harris LD,Briant JA,Sanders-Beer BE,Herbert R,Woodward R,Silvestri G,Pandrea I,Apetrei C,Hirsch VM,Brenchley JM
14-7179 was used in Flow cytometry/Cell sorting to demonstrate that coevolution with SIV has, in several cases, involved downregulation of receptors for the virus by otherwise-susceptible host target cells.
Thu Sep 01 00:00:00 EDT 2011
CD4-like immunological function by CD4- T cells in multiple natural hosts of simian immunodeficiency virus.
Journal of virology
Vinton C,Klatt NR,Harris LD,Briant JA,Sanders-Beer BE,Herbert R,Woodward R,Silvestri G,Pandrea I,Apetrei C,Hirsch VM,Brenchley JM
14-7179 was used in Flow cytometry/Cell sorting to demonstrate that coevolution with SIV has, in several cases, involved downregulation of receptors for the virus by otherwise-susceptible host target cells.
Thu Sep 01 00:00:00 EDT 2011
Loss of IL-17-producing CD8 T cells during late chronic stage of pathogenic simian immunodeficiency virus infection.
Journal of immunology (Baltimore, Md. : 1950)
Nigam P,Kwa S,Velu V,Amara RR
14-7179 was used in Flow cytometry/Cell sorting to characterise the anatomical distribution, phenotype, and functional quality of Tc17 and Th17 cells in SIV- and SIV+ rhesus macaques.
Sat Jan 15 00:00:00 EST 2011
Colorectal cancer-derived Foxp3(+) IL-17(+) T cells suppress tumour-specific CD8+ T cells.
Scandinavian journal of immunology
Ma C,Dong X
14-7179 was used in Neutralization to investigate the role of tumour-derived FoxP3(+), interleukin-17(+) T cells on suppressing tumour-specific CD8(+) T cells.
Fri Jul 01 00:00:00 EDT 2011
Local expression of interleukin-27 ameliorates collagen-induced arthritis.
Arthritis and rheumatism
Pickens SR,Chamberlain ND,Volin MV,Mandelin AM,Agrawal H,Matsui M,Yoshimoto T,Shahrara S
14-7179 was used in Flow cytometry/Cell sorting to explore the mechanism of action in mice with collagen-induced arthritis.
Mon Aug 01 00:00:00 EDT 2011
The anti-CD20 antibody rituximab reduces the Th17 cell response.
Arthritis and rheumatism
van de Veerdonk FL,Lauwerys B,Marijnissen RJ,Timmermans K,Di Padova F,Koenders MI,Gutierrez-Roelens I,Durez P,Netea MG,van der Meer JW,van den Berg WB,Joosten LA
14-7179-82 was used in Immunohistochemistry (Frozen) to demonstrate an unexpected role of B cells in the development of Th17 responses.
Wed Jun 01 00:00:00 EDT 2011
Chemoattractant receptors and lymphocyte egress from extralymphoid tissue: changing requirements during the course of inflammation.
Journal of immunology (Baltimore, Md. : 1950)
Brown MN,Fintushel SR,Lee MH,Jennrich S,Geherin SA,Hay JB,Butcher EC,Debes GF
14-7179 was used in Flow cytometry/Cell sorting to investigate the dynamics and mechanisms of T cell exit from inflamed tissues, showing that requirements change during the course of inflammation.
Fri Oct 15 00:00:00 EDT 2010
Transfer of tolerance to collagen type V suppresses T-helper-cell-17 lymphocyte-mediated acute lung transplant rejection.
Transplantation
Braun RK,Molitor-Dart M,Wigfield C,Xiang Z,Fain SB,Jankowska-Gan E,Seroogy CM,Burlingham WJ,Wilkes DS,Brand DD,Torrealba J,Love RB
14-7179 was used in Neutralization experiments to test whether Tregs from tolerant rats could suppress Th17-mediated rejection in the syngeneic model of lung transplantation.
Sun Dec 27 00:00:00 EST 2009
A monoclonal antibody selection for immunohistochemical examination of lymphoid tissues from non-human primates.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Kap YS,van Meurs M,van Driel N,Koopman G,Melief MJ,Brok HP,Laman JD,'t Hart BA
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Immunohistochemistry
Tue Dec 01 00:00:00 EST 2009
Interleukin-12 is the optimum cytokine to expand human Th17 cells in vitro.
Clinical and vaccine immunology : CVI
Nady S,Ignatz-Hoover J,Shata MT
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in ELISA
Mon Jun 01 00:00:00 EDT 2009
Interleukin-12 is the optimum cytokine to expand human Th17 cells in vitro.
Clinical and vaccine immunology : CVI
Nady S,Ignatz-Hoover J,Shata MT
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in ELISA
Mon Jun 01 00:00:00 EDT 2009
Prolonged, granulocyte-macrophage colony-stimulating factor-dependent, neutrophil survival following rheumatoid synovial fibroblast activation by IL-17 and TNFalpha.
Arthritis research & therapy
Parsonage G,Filer A,Bik M,Hardie D,Lax S,Howlett K,Church LD,Raza K,Wong SH,Trebilcock E,Scheel-Toellner D,Salmon M,Lord JM,Buckley CD
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Immunofluorescence
Tue Sep 23 00:00:00 EDT 2008
Distinct regulation of interleukin-17 in human T helper lymphocytes.
Arthritis and rheumatism
Chen Z,Tato CM,Muul L,Laurence A,O'Shea JJ
Published figure using IL-17A monoclonal antibody (Product # 14-7179-82) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2007