Search
Search
Invitrogen
Promotions
View available promotion(s)
Promo Code: RGLOW25 Save up to $900 and get the limited-edition holiday sweatshirt + matching beanie It’s been a year – this is your badge of honor for fluorescing through it all. Offer available until December 27.. Learn more
Spend Now, Save Big Later! Spend Now, Save Big Later! Make your New year's resolution to save: earn up to $3,500 off a future January offer when you purchase a qualifying amount before December 31st. Learn more
Description: This AB12-3G4 monoclonal antibody reacts with and inhibits the bioactivity of human IL-2. IL-2 is a 15 kDa cytokine expressed by B cells, T cells, and dendritic cells. IL-2 signals via a heterotrimeric receptor (CD25) and results in the activation of Jak/STAT pathways. IL-2 plays a critical role in the proliferation, activation, and differentiation of T cells. The development of CD4+CD25+ regulatory T cells is especially dependent on the presence of IL-2, making it an important immunoregulatory cytokine.
Applications Reported: This AB12-3G4 monoclonal antibody inhibits the bioactivity of human IL-2.
Applications Tested: The ND50 of AB12-3G4, as determined by the neutralization of Human IL-2-induced proliferation of the CTLL-2 cell line, is less than or equal to 60 ng/mL in the presence of 10 ng/mL Human IL-2 Recombinant Protein. The neutralization dose of this antibody will vary depending on cell type, assay method, and cytokine concentration.
Storage and handling: Use in a sterile environment.
Filtration: 0.2 µm post-manufacturing filtered.
Purity: Greater than 90%, as determined by SDS-PAGE.
Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.
Aggregation: Less than 10%, as determined by HPLC.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aldesleukin; H-IL-2; il 2; IL-2; ILN; Interleukin; Interleukin-2; Interleukin2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; M-IL-2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Gene Aliases: IL-2; IL2; lymphokine; TCGF
UniProt ID: (Human) P60568
Entrez Gene ID: (Human) 3558
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support