Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • IL-29 Antibodies

            Invitrogen

            IL-29 Monoclonal Antibody (C6)

            View all (15) IL-29 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite IL-29 Monoclonal Antibody (C6)

            • Antibody Testing Data (4)
            Application
            IL-29 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 4

            IL-29 Antibody (MA5-43734) in IHC (P)

            Immunohistochemistry of IL-29 in paraffin-embedded human cerebrum tissue. Samples were incubated in monoclonal IL-29 antibody (Product # MA5-43734) using a dilution of 10 µg/mL, followed by HRP-Linked Caprine Anti-Mouse IgG at a dilution of 2 µg/mL and DAB.
            View Product

            IL-29 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            IL-29 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            IL-29 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            IL-29 Antibody in Western Blot (WB)

            Product Details

            MA5-43734

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            0.5-2 µg/mL
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            5-40 µg/mL
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG2b, kappa

            Class

            Monoclonal

            Type

            Antibody

            Clone

            C6

            Immunogen

            Gly20-Thr200 (Accession Q8IU54), with N-terminal His Tag
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Protein A/G

            Storage buffer

            PBS, pH 7.4, with 50% glycerol

            Contains

            0.05% ProClin 300

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2912666

            Target Information

            IFN-lambdas constitute a novel family of interferons that mediate the induction of anti-viral protection in a wide variety of cells. There are three members in this family, namely lambda1, lambda2, and lambda3, which are also known as IL-29, IL-28A, and IL-28B, respectively. Similar to type I IFNs, IFN-lambdas activate an intracellular signaling to promote gene expression. IFN-lambdas mediate their anti-viral protection through a class II cytokine receptor complex composed of two essential receptor proteins, CRF2-12/IFN-lambdaR1, which is unique to IFN-lambdas, and CFR2-4/IL-10R2, which is shared with IL-10, IL-22, and IL-26 receptors. The IFN-lambdas, IL-28 and IL-29, have recently been reported to prime dendritic cells to induce proliferation of Foxp3-bearing regulatory T cells. DCs that differentiate in response to IFN-lambdas express high levels of class I and II MHC gene products, but low levels of costimulatory molecules. Moreover, these cells can specifically induce IL-2-dependent proliferation of CD4+CD25+FOXP3+ T cells with contact-dependent suppressive activity.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: Cytokine Zcyto21; FNL1; IFN lambda1; IFN-lambda-1; IL-29; ILN; Interferon lambda-1; Interferon lambda1; interferon, lambda 1; interferon-lambda-1; Interleukin; interleukin 29 (interferon, lambda 1); Interleukin-29

            View more View less

            Gene Aliases: IFNL1; IL-29; IL29; ZCYTO21

            View more View less

            UniProt ID: (Human) Q8IU54

            View more View less

            Entrez Gene ID: (Human) 282618

            View more View less

            Function(s)

            receptor binding cytokine activity interleukin-28 receptor binding interleukin superfamily

            Process(es)

            negative regulation of type 2 immune response JAK-STAT cascade negative regulation of cell proliferation negative regulation of interleukin-13 production negative regulation of interleukin-5 production positive regulation of interferon-gamma production positive regulation of tyrosine phosphorylation of STAT protein negative regulation of memory T cell differentiation innate immune response positive regulation of MHC class I biosynthetic process negative regulation of T cell differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of JAK-STAT cascade positive regulation of immune response defense response to virus
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0