Product References
Resveratrol Contrasts IL-6 Pro-Growth Effects and Promotes Autophagy-Mediated Cancer Cell Dormancy in 3D Ovarian Cancer: Role of miR-1305 and of Its Target ARH-I.
Cancers
Esposito A,Ferraresi A,Salwa A,Vidoni C,Dhanasekaran DN,Isidoro C
Published figure using IL-6 Receptor monoclonal antibody (Product # AHR0061) in Western Blot
Mon Apr 25 00:00:00 UTC 2022
Salivary Gland Stem Cells Age Prematurely in Primary Sjögren's Syndrome.
Arthritis & rheumatology (Hoboken, N.J.)
Pringle S,Wang X,Verstappen GMPJ,Terpstra JH,Zhang CK,He A,Patel V,Jones RE,Baird DM,Spijkervet FKL,Vissink A,Bootsma H,Coppes RP,Kroese FGM
AHR0061 was used in Immunohistochemistry (PFA fixed) to investigate the role of salivary gland stem cells in hyposalivation in primary SS.
Tue Jan 01 00:00:00 UTC 2019
Sorafenib inhibits signal transducer and activator of transcription-3 signaling in cholangiocarcinoma cells by activating the phosphatase shatterproof 2.
Hepatology (Baltimore, Md.)
Blechacz BR,Smoot RL,Bronk SF,Werneburg NW,Sirica AE,Gores GJ
Published figure using IL-6 Receptor monoclonal antibody (Product # AHR0061) in Western Blot
Tue Dec 01 00:00:00 UTC 2009
Involvement of hepcidin in the anemia of multiple myeloma.
Clinical cancer research : an official journal of the American Association for Cancer Research
Sharma S,Nemeth E,Chen YH,Goodnough J,Huston A,Roodman GD,Ganz T,Lichtenstein A
AHR0061 was used in blocking or activating experiment to investigate the role of hepcidin in the anemia of multiple myeloma
Sun Jun 01 00:00:00 UTC 2008
Interferon alpha induces cell death through interference with interleukin 6 signaling and inhibition of STAT3 activity.
Experimental cell research
Thyrell L,Arulampalam V,Hjortsberg L,Farnebo M,Grandér D,Pokrovskaja Tamm K
AHR0061 was used in blocking or activating experiment to establish that IFNalpha treatment results in reduced STAT3 activity in two myeloma cell lines, U266-1984 and U-1958
Thu Nov 15 00:00:00 UTC 2007
Retinoic acid inhibits IL-6-dependent but not constitutive STAT3 activation in Epstein-Barr virus-immortalized B lymphocytes.
International journal of oncology
Zancai P,Cariati R,Quaia M,Guidoboni M,Rizzo S,Boiocchi M,Dolcetti R
AHR0061 was used in flow cytometry to evaluate the effects of retinoic acid on IL-6 signaling in lymphoblastoid B-cell lines
Sun Aug 01 00:00:00 UTC 2004
Interleukin-6 enhances whereas tumor necrosis factor alpha and interferons inhibit integrin expression and adhesion of human mast cells to extracellular matrix proteins.
The Journal of investigative dermatology
Schoeler D,Grützkau A,Henz BM,Küchler J,Krüger-Krasagakis S
AHR0061 was used in flow cytometry to assess the effects of cytokines on integrin expression and mast cell adhesion.
Thu May 01 00:00:00 UTC 2003
Shedding of the interleukin-6 (IL-6) receptor (gp80) determines the ability of IL-6 to induce gp130 phosphorylation in human osteoblasts.
The Journal of biological chemistry
Vermes C,Jacobs JJ,Zhang J,Firneisz G,Roebuck KA,Glant TT
Fri May 10 00:00:00 UTC 2002
Human herpesvirus 8-encoded interleukin 6 activates HIV-1 in the U1 monocytic cell line.
AIDS (London, England)
Gage JR,Breen EC,Echeverri A,Magpantay L,Kishimoto T,Miles S,Martínez-Maza O
AHR0061 was used in ELISA to study human herpesvirus 8 interleukin 6.
Fri Oct 01 00:00:00 UTC 1999
Selective proteolytic cleavage of IL-2 receptor and IL-6 receptor ligand binding chains by neutrophil-derived serine proteases at foci of inflammation.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research
Bank U,Reinhold D,Schneemilch C,Kunz D,Synowitz HJ,Ansorge S
AHR0061 was used in flow cytometry to examine the role of role of neutrophil-derived serine proteases in traumatic brain injuries.
Mon Nov 01 00:00:00 UTC 1999