Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: RSPKSKRKGQ GRPGPLAPGP HQVPLDLVSR MKPYARMEEY ERNIEEMVAQ LRNSSELAQR KCEVNLQLWM SNKRSLSPWG YSINHDPSRI PVDLPEARCL
The protein encoded by this gene is a T cell-derived cytokine that shares sequence similarity with IL17. This cytokine was reported to stimulate the release of TNF alpha (TNF) and IL1 beta (IL1B) from a monocytic cell line. Immunohistochemical analysis of several nerve tissues indicated that this cytokine is primarily localized to neuronal cell bodies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytokine Zcyto7; cytokine-like protein ZCYTO7; IL-17B; IL-20; interleukin 20; interleukin-17 beta; Interleukin-17B; Interleukin-20; Interleukin20; neuronal interleukin-17 related factor; Neuronal interleukin-17-related factor
Gene Aliases: IL-17B; IL-20; IL17B; IL20; NIRF; UNQ516/PRO1031; ZCYTO7
UniProt ID: (Human) Q9UHF5
Entrez Gene ID: (Human) 27190
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support