Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: Human lung leukocytes liver or thymus. spleen
Cellular Location: Membrane.
IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL-18R-1; IL-18R-alpha; IL-18R1; IL-1Rrp; IL1 receptor-related protein; IL18Ralpha2; IL1R-rp; Interleukin-18 receptor 1; Interleukin-18 receptor alpha
Gene Aliases: CD218a; CDw218a; IL-1Rrp; IL18R1; IL18RA; IL1RRP
UniProt ID: (Human) Q13478
Entrez Gene ID: (Human) 8809
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support