Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DGCKDIFMKN EILSASQPFA FNCTFPPITS GEVSVTWYKN SSKIPVSKII QSRIHQDETW ILFLPMEWGD SGVYQCVIKG RDSCHRIHVN LTVFEKHWC
The protein encoded by this gene is a member of the interleukin 1 receptor family. An experiment with transient gene expression demonstrated that this receptor was incapable of binding to interleukin 1 alpha and interleukin 1 beta with high affinity. This gene and four other interleukin 1 receptor family genes, including interleukin 1 receptor, type I (IL1R1), interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 1 (IL1RL1), and interleukin 18 receptor 1 (IL18R1), form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IL-1 receptor-related protein 2; IL-1Rrp2; IL-36 receptor; IL-36R; interleukin 1 receptor-like 2; Interleukin-1 receptor-like 2; Interleukin-1 receptor-related protein 2
Gene Aliases: IL-1Rrp2; IL-36R; IL1R-rp2; IL1RL2; IL1RRP2
UniProt ID: (Human) Q9HB29
Entrez Gene ID: (Human) 8808
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support