Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Orangutan
IMPA1, also known as Myo-inositol monophosphatase 1, is responsible for the procurement of inositol that is required for synthesis of phosphatidylinositol and polyphosphoinositides. IMPA1 exists as a homodimer and has been identified as the pharmacological target for lithium action in the brain. IMPA1 is the principal enzyme of the phosphatidyl inositol signaling pathway, and inhibition of inositol monophosphatase hydrolysis may underlie the anti-manic and anti-depressant actions of Li+. Studies indicate that a variation in the 277 codon coding region of the IMPA1 gene has not been observed in manic-depressive patients, therefore suggesting that polymorphisms or mutations in the noncoding regions of this gene may influence the lithium response in psychiatric patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-galactose 1-phosphate phosphatase; IMP 1; IMPase 1; Inositol monophosphatase 1; inositol(myo)-1(or 4)-monophosphatase 1; Inositol-1(or 4)-monophosphatase 1; Lithium-sensitive myo-inositol monophosphatase A1; myo-inositol monophosphatase 1; testicular tissue protein Li 94
Gene Aliases: IMP; IMPA; IMPA1
UniProt ID: (Human) P29218
Entrez Gene ID: (Human) 3612
Molecular Function: phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support