Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GRFSLFGLG GEPGGGAAEP AAAADGGTVD LREMLAVSVL AAVRGGDEVR RVRESNVLHE KSKGKTREGA EDKMTSGDVL SNRKMFYLLK AAFPSVQINT EEHVDAADQE VILWDHKIPE DILEEVTTPK EVPAESVTVW IDPLDATQEY TEDLRKYVTT MVCVAVNGKP MLGVIHKPFS EYTAWAMVDG GSNVKARSSY NEKTPRIVVS RSHSGMVKQV ALQTFGNQTT IIPAGGAGYK VLALLDVPDK SQEKADLYIH VTYIKKWDIC AGNAILKALG GHMTTLSGEE ISYTGSDGIE GGLLASIRMN HQALVRKLPD LEKTGHK (34-359 aa encoded by BC067814)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'(2'), 5'-bisphosphate nucleotidase 2; EC 3.1.3; EC 3.1.3.25; FLJ20421; Golgi 3-prime phosphoadenosine 5-prime phosphate 3-prime phosphatase; Golgi-resident adenosine 3',5'-bisphosphate 3'-phosphatase; golgi-resident nucleotide phosphatase; Golgi-resident PAP phosphatase; IMP 3; IMP3; IMPA3IMPase 3; IMPase 3; inositol monophosphatase 3; Inositol monophosphatase domain-containing protein 1; inositol-1(or 4)-monophosphatase 3; Myo-inositol monophosphatase A3; Phosphoadenosine phosphate 3'-nucleotidase
Gene Aliases: 1110001C20Rik; AA408880; AI451589; AL022796; B230207P20; BPNT2; GPAPP; IMP 3; IMP-3; IMPA3; IMPAD1; Jaws
UniProt ID: (Human) Q9NX62, (Mouse) Q80V26
Entrez Gene ID: (Human) 54928, (Mouse) 242291
Molecular Function: phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support