Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'(2'), 5'-bisphosphate nucleotidase 2; Ac2-190; EC 3.1.3; EC 3.1.3.25; FLJ20421; Golgi 3-prime phosphoadenosine 5-prime phosphate 3-prime phosphatase; Golgi-resident adenosine 3',5'-bisphosphate 3'-phosphatase; golgi-resident nucleotide phosphatase; Golgi-resident PAP phosphatase; IMP 3; IMP3; IMPA3IMPase 3; IMPase 3; inositol monophosphatase 3; Inositol monophosphatase domain-containing protein 1; inositol-1(or 4)-monophosphatase 3; Myo-inositol monophosphatase A3; Phosphoadenosine phosphate 3'-nucleotidase
Gene Aliases: 1110001C20Rik; AA408880; AI451589; AL022796; B230207P20; BPNT2; GPAPP; IMP 3; IMP-3; IMPA3; IMPAD1; Jaws; RGD1306455
UniProt ID: (Human) Q9NX62, (Mouse) Q80V26, (Rat) D4AD37
Entrez Gene ID: (Human) 54928, (Mouse) 242291, (Rat) 312952
Molecular Function: phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support