Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji.
Predicted reactivity: Mouse (95%), Rat (82%), Xenopus laevis (88%), Bovine (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene acts as a homotetramer to regulate cell growth. The encoded protein is an enzyme that catalyzes the synthesis of xanthine monophosphate (XMP) from inosine-5'-monophosphate (IMP). This is the rate-limiting step in the de novo synthesis of guanine nucleotides. Defects in this gene are a cause of retinitis pigmentosa type 10 (RP10). Several transcript variants encoding different isoforms have been found for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781N0678; IMP (inosine 5'-monophosphate) dehydrogenase 1; IMP (inosine monophosphate) dehydrogenase 1; IMP dehydrogenase 1; IMPD 1; IMPDH 1; IMPDH-I; Inosine-5'-monophosphate dehydrogenase 1
Gene Aliases: IMPD; IMPD1; IMPDH-I; IMPDH1; LCA11; RP10; sWSS2608
UniProt ID: (Human) P20839
Entrez Gene ID: (Human) 3614
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support