Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MADYLISGGT SYVPDDGLTA QQLFNCGDGL TYNDFLILPG YIDFTADQVD LTSALTKKIT LKTPLVSSPM DTVTEAGMAI AMALTGGIGF IHHNCTPEFQ ANEVRKVKKY EQGFITDPVV LSPKDRVRDV FEAKARHGFC GIPITDTGRM GSRLVGIISS RDIDFLKEEE HDCFLEEIMT KREDLVVAPA GITLKEANEI LQRSKKGKLP IVNEDDELVA IIARTDLKKN RDYPLASKDA KKQLLCGAAI GTHEDDKYRL DLLAQAGVDV VVLDSSQGNS IFQINMIKYI KDKYPNLQVI GGNVVTAAQA KNLIDAGVDA LRVGMGSGSI CITQEVLACG RPQATAVYKV SEYARRFGVP VIADGGIQNV GHIAKALALG ASTVMMGSLL AATTEAPGEY FFSDGIRLKK YRGMGSLDAM DKHLSSQNRY FSEADKIKVA QGVSGAVQDK GSIHKFVPYL IAGIQHSCQD IGAKSLTQVR AMMYSGELKF EKRTSSAQVE GGVHSLHSYE KRLF
This gene encodes the rate-limiting enzyme in the de novo guanine nucleotide biosynthesis. It is thus involved in maintaining cellular guanine deoxy- and ribonucleotide pools needed for DNA and RNA synthesis. The encoded protein catalyzes the NAD-dependent oxidation of inosine-5'-monophosphate into xanthine-5'-monophosphate, which is then converted into guanosine-5'-monophosphate. This gene is up-regulated in some neoplasms, suggesting it may play a role in malignant transformation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IMP (inosine 5'-monophosphate) dehydrogenase 2; IMP (inosine monophosphate) dehydrogenase 2; IMP dehydrogenase 2; IMP dehydrogenase II; IMP oxireductase 2; IMPD 2; IMPDH 2; IMPDH II; IMPDH2; inosine 5' phosphate dehydrogenase 2; inosine monophosphate dehydrogenase type II; Inosine-5'-monophosphate dehydrogenase 2; Inosine-5'-monophosphate dehydrogenase type II
Gene Aliases: IMPD2; IMPDH-II; IMPDH2
UniProt ID: (Human) P12268
Entrez Gene ID: (Human) 3615
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support