- Order Status
- Quick Order
-
Don't have an account ? Create Account
Immunogen sequence: ERRNTGYQFI YYSPENTAKA KEVLSNINQL QPLIATHADL LLNSASQHSP DSLKNSLKML SEKTELFVHA FKDQLVRSAL LALYTA
Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
Catalyzes the hydrolysis of the 4-position phosphate of phosphatidylinositol 3,4-bisphosphate, inositol 1,3,4- trisphosphate and inositol 1,4-bisphosphate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: inositol polyphosphate 4-phosphatase II; 4-phosphatase II; Inositol polyphosphate 4-phosphatase type II; inositol polyphosphate-4-phosphatase, type II, 105kD; inositol polyphosphate-4-phosphatase, type II, 105kDa; Type II inositol 3,4-bisphosphate 4-phosphatase; type II inositol-3,4-bisphosphate 4-phosphatase
Gene Aliases: INPP4B
UniProt ID: (Human) O15327
Entrez Gene ID: (Human) 8821
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support