Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MELFQAKDHY ILQQGERALW CSRRDGGLQL RPATDLLLAW NPICLGLVEG VIGKIQLHSD LPWWLILIRQ KALVGKLPGD HEVCKVTKIA VLSLSEMEPQ DLELELCKKH HFGINKPEKI IPSPDDSKFL LKTFTHIKSN VSAPNKKKVK ESKEKEKLER RLLEELLKMF MDSESFYYSL TYDLTNSVQR QSTGERDGRP LWQKVDDRFF WNKYMIQDLT EIGTPDVDFW IIPMIQGFVQ IEELVVNYTE SSDDEKSSPE TPPQESTCVD DIHPRFLVAL ISRRSRHRAG MRYKRRGVDK NGNVANYVET EQLIHVHNHT LSFVQTRGSV PVFWSQVGYR YNPRPRLDRS EKETVAYFCA HFEEQLNIYK KQVIINLVDQ AGREKIIGDA YLKQVLLFNN SHLTYVSFDF HEHCRGMKFE NVQTLTDAIY DIILDMKWCW VDEAGVICKQ EGIFRVNCMD CLDRTNVVQA AIARVVMEQQ LKKLGVMPPE QPLPVKCNRI YQIMWANNGD SISRQYAGTA ALKGDFTRTG ERKLAGVMKD GVNSANRYYL NRFKDAYRQA VIDLMQGIPV TEDLYSIFTK EKEHEALHKE NQRSHQELIS QLLQSYMKLL LPDDEKFHGG WALIDCDPSL IDATHRDVDV LLLLSNSAYY VAYYDDEVDK VNQYQRLSLE NLEKIEIGPE PTLFGKPKFS CMRLHYRYKE ASGYFHTLRA VMRNPEEDGK DTLQCIAEML QITKQAMGSD LPIIEKKLER KSSKPHEDII GIRSQNQGSL AQGKNFLMSK FSSLNQKVKQ TKSNVNIGNL RKLGNFTKPE MKVNFLKPNL KVNLWKSDSS LETMENTGVM DKVQAESDGD MSSDNDSYHS DEFLTNSKSD EDRQLANSLE SVGPIDYVLP SCGIIASAPR LGSRSQSLSS TDSSVHAPSE ITVAHGSGLG KGQESPLKKS PSAGDVHILT GFAKPMDIYC HRFVQDAQNK VTHLSETRSV SQQASQERNQ MTNQVSNETQ SESTEQTPSR PSQLDVSLSA TGPQFLSVEP AHSVASQKTP TSASSMLELE TGLHVTPSPS ESSSSRAVSP FAKIRSSMVQ VASITQAGLT HGINFAVSKV QKSPPEPEII NQVQQNELKK MFIQCQTRII QI
SAC2 is an inositol 1,4,5-trisphosphate (InsP3) 5-phosphatase and contains a Sac domain. The activity of this protein is specific for phosphatidylinositol 4,5-bisphosphate and phosphatidylinositol 3,4,5-trisphosphate. Alternatively spliced transcript variants have been observed, but most of them are not thought to be protein-coding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 3.1.3.36; hSAC2; Inositol polyphosphate 5-phosphatase F; OCRL 1; Phosphatidylinositide phosphatase SAC2; Sac domain-containing inositol phosphatase 2; Sac domain-containing phosphoinositide 4-phosphatase 2; sac domain-containing phosphoinositide 5-phosphatase 2
Gene Aliases: hSAC2; INPP5F; KIAA0966; MSTP007; MSTP047; MSTPO47; SAC2
UniProt ID: (Human) Q9NT51
Entrez Gene ID: (Human) 22876
Molecular Function: phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support