Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
SAC2 is an inositol 1,4,5-trisphosphate (InsP3) 5-phosphatase and contains a Sac domain. The activity of this protein is specific for phosphatidylinositol 4,5-bisphosphate and phosphatidylinositol 3,4,5-trisphosphate. Alternatively spliced transcript variants have been observed, but most of them are not thought to be protein-coding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 3.1.3.36; hSAC2; Inositol polyphosphate 5-phosphatase F; OCRL 1; Phosphatidylinositide phosphatase SAC2; Sac domain-containing inositol phosphatase 2; Sac domain-containing phosphoinositide 4-phosphatase 2; sac domain-containing phosphoinositide 5-phosphatase 2
Gene Aliases: hSAC2; INPP5F; KIAA0966; MSTP007; MSTP047; MSTPO47; SAC2
UniProt ID: (Human) Q9NT51
Entrez Gene ID: (Human) 22876
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support