Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: LECLYQYID SAGLSSVPLY FISPVANSSL EFSQIFAEWL CHNKQSKVYL PEPPFPHAEL IQTNKLKHYP SIHGDFSNDF RQPCVVFTGH PSLRFGDVVH FMELWGKSSL NTVIFTEPDF SYLEALAPYQ PLAMKCIYCP IDTRLNFIQV SKLLKEVQPL HVVCPEQYTQ PPPAQSHRMD LMIDCQPPAM SYRRAEVLAL PFKRRYEKIE IMPELADSLV PMEIKPGISL ATVSAVLHTK DNKHLLQPPP RPAQPTSGKK RKRVSDDVPD CKVLKPLLSG SIPVEQFVQT LEKHGFSDIK VEDTAKGHIV LLQEAETLIQ IEEDSTHIIC DNDEMLRVRL RDLVLKFLQK F (309-658 aa encoded by BC025267)
INTS9 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A)and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1)and U2 (RNU2)(Baillat et al., 2005).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Int9; Integrator complex subunit 9; Protein related to CPSF subunits of 74 kDa; RC-74
Gene Aliases: CPSF2L; INT9; INTS9; RC74
UniProt ID: (Human) Q9NV88
Entrez Gene ID: (Human) 55756
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support