Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • IP3 Receptor 2 Antibodies

            Invitrogen

            IP3 Receptor 2 Polyclonal Antibody

            5 Published Figures
            13 References
            View all (11) IP3 Receptor 2 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # PA1-904
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (EUR)
            402,65
            Online Exclusive
            536,00
            Save 133,35 (25%)
            100 µL
             
            Check your price
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite IP3 Receptor 2 Polyclonal Antibody

            • Antibody Testing Data (2)
            • Published Figures (5)
            Application
            IP3 Receptor 2 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 7

            IP3 Receptor 2 Antibody (PA1-904) in ICC/IF

            Immunofluorescence analysis of IP3 Receptor II was done on 70% confluent log phase SH-SY5Y cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with IP3 Receptor II Rabbit Polyclonal Antibody (Product # PA1-904) at 2 µg/mL in 0.1% BSA and incubated for 3 hours ... View More
            View Product

            IP3 Receptor 2 Antibody in Immunocytochemistry (ICC/IF)
            IP3 Receptor 2 Antibody in Immunohistochemistry (IHC)
            IP3 Receptor 2 Antibody in Immunocytochemistry (ICC/IF)
            IP3 Receptor 2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
            IP3 Receptor 2 Antibody in Immunohistochemistry (IHC)
            IP3 Receptor 2 Antibody in Immunohistochemistry (IHC)
            IP3 Receptor 2 Antibody in Immunohistochemistry (IHC)

            Product Details

            PA1-904

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            -
            View 2 publications 2 publications

            Immunohistochemistry (IHC)

            -
            View 3 publications 3 publications

            Immunohistochemistry (Frozen) (IHC (F))

            1:100
            -

            Immunocytochemistry (ICC/IF)

            1:100
            View 8 publications 8 publications
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Amphibian, Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Synthetic Peptide: E(317) L N P D Y R D A Q N E G K T V R D(334).
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS with 1mg/mL BSA

            Contains

            0.05% sodium azide

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2130127

            Product Specific Information

            PA1-904 detects inositol 1,4,5-trisphosphate receptor type-II (IP3R-II) from rat and mouse tissues.

            PA1-904 has been successfully used in immunohistochemistry, immunofluorescence and immunocytochemistry procedures. Immunohistochemical staining of IP3R-II in rat brain with PA1-904 results in staining of the hippocampus, corpus collosum, and cerebellum. This product fails to detect IP3R-II in Western blot procedures.

            PA1-904 immunizing peptide corresponds to amino acid residues 317-334 from rat IP3R-II. PA1-904 immunizing peptide (Cat. # PEP-024) is available for use in neutralization and control experiments.

            Target Information

            Inositol 1,4,5-trisphosphate (IP3) is a second messenger for many growth factors, hormones, and neuro-transmitters. Upon binding to IP3R, IP3 triggers the release of intracellular, luminal calcium to the cytosol. Functional IP3R is a homo- or heterotetramer of ~240 kDa glycoprotein subunits. IP3R protein is structurally and functionally related to one other important intracellular calcium-release channel, the ryanodine receptor. The similarity between the two receptors continues at the physiological level owing to a physical association each receptor can have with the immunophilin protein, FKBP12. Phosphorylation of the IP3R by PKC causes an increase in IP3-mediated calcium release. Concomitantly, the phosphatase activity of calcineurin is stimulated upon its association with the FKBP12-IP3R complex. Calcium release is reduced when the PKC target site on the IP3R is dephosphorylated by calcineurin resulting in calcium oscillations. Mammalian IP3R subunits are the product of three distinct genes that are widely expressed and differentially regulated. IP3R type I (IP3R-I) has been detected in heart, liver, kidney, ovary, and Purkinje neurons of the cerebellum. IP3R-II is found predominantly in the brain. IP3R-III is known to be expressed in pancreatic islets, kidney, and the gastrointestinal tract.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: cilia and flagella associated protein 48; inositol 1,4,5-triphosphate receptor 2; inositol 1,4,5-triphosphate receptor 5; inositol 1,4,5-triphosphate receptor, type 2; Inositol 1,4,5-trisphosphate receptor type 2; Inositol 1,4,5-trisphosphate type V receptor; Inositol 1,4,5-trisphosphate-gated calcium channel ITPR2; inositol triphosphate receptor type 2; inositol trisphosphate receptor type 2; insP3R2; IP3 receptor; IP3 receptor isoform 2; IP3R; IP3R 2; Type 2 inositol 1,4,5-trisphosphate receptor; Type 2 InsP3 receptor

            View more View less

            Gene Aliases: AI649341; ANHD; CFAP48; InsP3R-2; InsP3R-5; INSP3R2; IP3R2; ITPR2; Itpr5

            View more View less

            UniProt ID: (Human) Q14571, (Mouse) Q9Z329, (Rat) P29995

            View more View less

            Entrez Gene ID: (Human) 3709, (Mouse) 16439, (Rat) 81678

            View more View less

            Function(s)

            inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity calcium ion transmembrane transporter activity phosphatidylinositol binding ion channel activity calcium channel activity calcium ion binding calcium-release channel activity inositol 1,4,5 trisphosphate binding ligand-gated ion channel

            Process(es)

            response to hypoxia transport signal transduction platelet activation inositol phosphate-mediated signaling regulation of insulin secretion release of sequestered calcium ion into cytosol cellular response to cAMP cellular response to ethanol regulation of cardiac conduction ion transport calcium ion transport transmembrane transport calcium ion transmembrane transport cellular response to hypoxia
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            • Price & Freight Policy
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • German Supply Chain Due Diligence Act
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Legal Notice
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            germany | deutschland flag icon
            Germany | Deutschland

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-7qv8r:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0