Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: MCVCQTMEVG QYGKNASRAG DRGVLLEPFI HQVGGHSSMM RYDDHTVCKP LISREQRFYE SLPPEMKEFT PEYKGVVSVC FEGDSDGYIN LVAYPYVESE TVEQDDTTER EQPRRKHSRR SLHRSGSGSD HKEEKASLSL ETSESSQEAK SPKVELHSHS EVPFQMLDGN SGLSSEKISH NPWSLRCHKQ QLSRMRSESK DRKLYKFLLL ENVVHHFKYP
This gene encodes a member of the inositol phosphokinase family. The encoded protein may be responsible for the conversion of inositol hexakisphosphate (InsP6) to diphosphoinositol pentakisphosphate (InsP7/PP-InsP5). It may also convert 1,3,4,5,6-pentakisphosphate (InsP5) to PP-InsP4. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP:1D-myo-inositol-hexakisphosphate phosphotransferase; Inositol hexakisphosphate kinase 1; inositol hexakisphosphate kinase 6; Inositol hexaphosphate kinase 1; InsP6 kinase 1; Pi uptake stimulator
Gene Aliases: 1200016D08Rik; AU022776; IHPK1; InsP6; InsP6k1; IP6K1; Itpk6; KIAA0263; PiUS
UniProt ID: (Human) Q92551, (Rat) Q9ESM0, (Mouse) Q6PD10
Entrez Gene ID: (Human) 9807, (Rat) 50560, (Mouse) 27399
Molecular Function:
kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support