Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Raji, mouse brain.
Predicted reactivity: Mouse (91%), Rat (90%), Bovine (92%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the inositol phosphokinase family. The encoded protein has 3-kinase, 5-kinase and 6-kinase activities on phosphorylated inositol substrates. The encoded protein plays an important role in the biosynthesis of inositol 1, 3, 4, 5, 6-pentakisphosphate, and has a preferred 5-kinase activity. This gene may play a role in nuclear mRNA export. Pseudogenes of this gene are located on the long arm of chromosome 13 and the short arm of chromosome 19.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inositol 1,3,4,6-tetrakisphosphate 5-kinase; Inositol polyphosphate multikinase
Gene Aliases: 2410017C19Rik; AA408208; IMPK; Ipk2; IPMK
UniProt ID: (Human) Q8NFU5, (Mouse) Q7TT16
Entrez Gene ID: (Human) 253430, (Mouse) 69718
Molecular Function:
kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support