Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene encodes a member of the IQGAP family. The protein contains four IQ domains, one calponin homology domain, one Ras-GAP domain and one WW domain. It interacts with components of the cytoskeleton, with cell adhesion molecules, and with several signaling molecules to regulate cell morphology and motility. Expression of the protein is upregulated by gene amplification in two gastric cancer cell lines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cdc42-Rac1 effector protein; DISP2; DISPB; p195; Ras GTPase-activating-like protein IQGAP1; RasGAP-like with IQ motifs
Gene Aliases: AA682088; D7Ertd237e; D7Ertd257e; HUMORFA01; IQGAP1; KIAA0051; mKIAA0051; p195; SAR1
UniProt ID: (Human) P46940, (Mouse) Q9JKF1
Entrez Gene ID: (Human) 8826, (Mouse) 29875, (Rat) 361598
Molecular Function: G-protein modulator protein-binding activity modulator GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support