Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A recommended positive control is A-20 cell lysate.
At least four isoforms of IQSEC1 are known to exist. IQSEC1 antibody is not expected to cross-react with IQSEC2.
ADP ribosylation factors (Arfs) are small GTP-binding proteins known for their role in vesicular transport, where they nucleate the assembly of coat protein complexes at sites of carrier vesicle formation. IQSEC1 (GEP100/BRAG2), a guanine nucleotide exchange protein for ARF6, belongs to the BRAG family and contains one IQ domain, one PH domain and one SEC7 domain. As a known activator of ARF6, IQSEC1 regulates cell surface levels of Beta1 integrin and its knockdown blocked tumour metastasis. IQSEC1 links EGFR signaling to ARF6 activation and may contribute to the cancer metastasis and malignancy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factors guanine nucleotide-exchange protein 100; ADP-ribosylation factors guanine nucleotide-exchange protein 2; BRAG2; brefeldin A-resistant ARF-GEF2; Brefeldin-resistant Arf-GEF 2 protein; IQ motif and Sec7 domain 1; IQ motif and SEC7 domain-containing protein 1
Gene Aliases: ARF-GEP100; ARFGEP100; AW561907; BC026481; BRAG2; cI-43; D6Ertd349e; GEP100; IQSEC1; KIAA0763
UniProt ID: (Human) Q6DN90, (Mouse) Q8R0S2
Entrez Gene ID: (Human) 9922, (Mouse) 232227, (Rat) 686590
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support