Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MSELYPLQIS PVSSYAESET TDSVPSDEES AEGRPHWRKR NIEGKQYLSN MGTRGSYLLP GMASFVTSNK PDLQVTIKEE SNPVPYNSSW PPFQDLPLSS
IRF2 belongs to the interferon regulatory factor family. IRF2 regulates the transcription regulation important to cell proliferation as well as immune responses by binding to the upstream regulatory region of type I IFN and IFNinducible MHC class I genes (the interferon consensus sequence (ICS)) and repression of those genes. However, IRF2 also functions as a transcriptional activator of histone H4 by interactions with NF-kappa B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686F0244; Interferon regulatory factor 2; IRF-2; OTTHUMP00000218576; OTTHUMP00000218580
Gene Aliases: IRF-2; IRF2
UniProt ID: (Human) P14316
Entrez Gene ID: (Human) 3660
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support