Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: TPPPFEIFFC FGEEWPDRKP REKKLITVQV VPVAARLLLE MFSGELSWSA DSIRLQISNP DLKDRMVEQF KELHHIWQSQ QRLQPVAQAP PGAGLGVGQG PWPMHPAGMQ
This gene encodes a member of the interferon regulatory factor (IRF) family, a group of transcription factors with diverse roles, including virus-mediated activation of interferon, and modulation of cell growth, differentiation, apoptosis, and immune system activity. Members of the IRF family are characterized by a conserved N-terminal DNA-binding domain containing tryptophan (W) repeats. Alternative splice variants encoding different isoforms exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Interferon regulatory factor 5; interferon regulatory factor 5 bone marrow; IRF-5; OTTHUMP00000212907
Gene Aliases: AW491843; IRF5; mirf5; SLEB10
UniProt ID: (Human) A4D1J8, (Mouse) P56477
Entrez Gene ID: (Human) 3663, (Mouse) 27056
Molecular Function: winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support