Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: DPKDAASKPS GEGSFSKPGD GGSPSKPSDH EPPKNKAKRP NRLSFITKGY KIKPKPQKPT HEQREADSSS DYVNMDFTKR ESNTPAPSTQ GLPDSWGIIA E
IRS4 encodes the insulin receptor substrate 4, a cytoplasmic protein that contains many potential tyrosine and serine/threonine phosphorylation sites. Tyrosine-phosphorylated IRS4 protein has been shown to associate with cytoplasmic signalling molecules that contain SH2 domains. The IRS4 protein is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 160 kDa phosphotyrosine protein; Insulin receptor substrate 4; IRS-4; Phosphoprotein of 160 kDa; pp160; py160; RP23-415G1.3
Gene Aliases: IRS-4; IRS4; PY160
UniProt ID: (Human) O14654
Entrez Gene ID: (Human) 8471
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support