Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
The protein encoded by this gene has been identified as a brain-specific angiogenesis inhibitor (BAI1)-binding protein. This adaptor protein links membrane bound G-proteins to cytoplasmic effector proteins. This protein functions as an insulin receptor tyrosine kinase substrate and suggests a role for insulin in the central nervous system. It also associates with a downstream effector of Rho small G proteins, which is associated with the formation of stress fibers and cytokinesis. This protein is involved in lamellipodia and filopodia formation in motile cells and may affect neuronal growth-cone guidance. This protein has also been identified as interacting with the dentatorubral-pallidoluysian atrophy gene, which is associated with an autosomal dominant neurodegenerative disease. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAI-associated protein 2; BAI1-associated protein 2; BAR/IMD domain-containing adapter protein 2; Brain-specific angiogenesis inhibitor 1-associated protein 2; Fas ligand-associated factor 3; FLAF3; insulin receptor substrate 53; insulin receptor substrate of 53 kDa; insulin receptor substrate p53; Insulin receptor substrate p53/p58; Insulin receptor substrate protein of 53 kDa; Insulin receptor tyrosine kinase 53 kDa substrate; Insulin receptor tyrosine kinase substrate protein p53; IRS-58; IRSp53; IRSp53/58
Gene Aliases: BAIAP2; BAP2; FLAF3; IRSP53; R75030
UniProt ID: (Human) Q9UQB8, (Mouse) Q8BKX1, (Rat) Q6GMN2
Entrez Gene ID: (Human) 10458, (Mouse) 108100, (Rat) 117542
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support