Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAAWGSSLT AATQRAVTPW PRGRLLTASL GPQARREASS SSPEAGEGQI CLTDSCVQRL LEITEGSEFL RLQVEGGGCS GFQYKFSLDT VINPDDRVFE QGGARVVVDS DSLAFVKGAQ VDFSQELIRS SFQVLNNPQA QQGCSCGSSF SIKL
ISCA2 is involved in the assembly of mitochondrial iron-sulfur proteins. Probably involved in the binding of an intermediate of Fe/S cluster assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HESB-like domain-containing protein 1; Iron-sulfur cluster assembly 2 homolog, mitochondrial
Gene Aliases: c14_5557; HBLD1; ISA2; ISCA2; MMDS4
UniProt ID: (Human) Q86U28
Entrez Gene ID: (Human) 122961
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support