Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
Recommended positive controls: Raji, mouse spleen.
Predicted reactivity: Mouse (83%), Rat (82%), Pig (81%), Rhesus Monkey (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Interferon-induced antiviral exoribonuclease that acts on single-stranded RNA and also has minor activity towards single-stranded DNA. Exhibits antiviral activity against RNA viruses including hepatitis C virus (HCV), hepatitis A virus (HAV) and yellow fever virus (YFV) in an exonuclease-dependent manner. May also play additional roles in the maturation of snRNAs and rRNAs, and in ribosome biogenesis. [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Estrogen-regulated transcript 45 protein; interferon stimulated exonuclease; interferon stimulated exonuclease gene 20kDa; Interferon-stimulated gene 20 kDa protein; Promyelocytic leukemia nuclear body-associated protein ISG20; Protein DnaQL
Gene Aliases: 1600023I01Rik; 2010107M23Rik; 20kDa; CD25; DnaQL; HEM45; ISG20
UniProt ID: (Human) Q96AZ6, (Mouse) Q9JL16
Entrez Gene ID: (Human) 3669, (Mouse) 57444
Molecular Function:
exoribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support