Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MPRAPAPLYA CLLGLCALLP RLAGLNICTS GSATSCEECL LIHPKCAWCS KEDFGSPRSI TSRCDLRANL VKNGCGGEIE SPASSFHVLR SLPLSSKGSG SAGWDVIQMT PQEIAVNLRP GDKTTFQLQV RQVEDYPVDL YYLMDLSLSM KDDLDNIRSL GTKLAEEMRK LTSNFRLGFG SFVDKDISPF SYTAPRYQTN PCIGYKLFPN CVPSFGFRHL LPLTDRVDSF NEEVRKQRVS RNRDAPEGGF DAVLQAAVCK EKIGWRKDAL HLLVFTTDDV PHIALDGKLG GLVQPHDGQC HLNEANEYTA SNQMDYPSLA LLGEKLAENN INLIFAVTKN HYMLYKNFTA LIPGTTVEIL DGDSKNIIQL IINAYNSIRS KVELSVWDQP EDLNLFFTAT CQDGVSYPGQ RKCEGLKIGD TASFEVSLEA RSCPSRHTEH VFALRPVGFR DSLEVGVTYN CTCGCSVGLE PNSARCNGSG TYVCGLCECS PGYLGTRCEC QDGENQSVYQ NLCREAEGKP LCSGRGDCSC NQCSCFESEF GKIYGPFCEC DNFSCARNKG VLCSGHGECH CGECKCHAGY IGDNCNCSTD ISTCRGRDGQ ICSERGHCLC GQCQCTEPGA FGEMCEKCPT CPDACSTKRD CVECLLLHSG KPDNQTCHSL CRDEVITWVD TIVKDDQEAV LCFYKTAKDC VMMFTYVELP SGKSNLTVLR EPECGNTPNA MTILLAVVGS ILLVGLALLA IWKLLVTIHD RREFAKFQSE RSRARYEMAS NPLYRKPIST HTVDFTFNKF NKSYNGTVD
Integrins are important extracellular matrix (ECM) receptor proteins located on cell surfaces. They are hetrodimers composed of an alpha and a beta transmembrane glycoprotein subunit. Around 22 different integrins (different alpha/ beta subunit combinations) are found in nature. Integrins are generally present in high concentrations at the cell surface, but, unlike most other cell-surface receptors, they bind ligands with very low affinity. Due to their weak individual binding, integrins need to cluster and bind in-groups in order to effectively bind the ECM. Integrins bind many different ligands including laminin. Each integrin is made up of a large N-terminal extracellular domain that binds the ECM ligand and a small C-terminal cytoplasmic domain that mediates interaction with the actin cytoskeleton and signaling function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ26658; GP3A; integrin beta 5; Integrin beta-5; integrin, beta 5; testis secretory sperm-binding protein Li 217p
Gene Aliases: [b]-5; [b]5; [b]5A; [b]5B; AA475909; AI874634; beta-5; beta5; ESTM23; ITGB5
UniProt ID: (Human) P18084, (Mouse) O70309
Entrez Gene ID: (Human) 3693, (Mouse) 16419
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support