Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 5
PA1-1755 detects HC1 from human and mouse samples.
PA1-1755 has been successfully used in immunofluorescence and immunohistochemical staining of paraffin embedded tissue sections.
The PA1-1755 immunogen is a synthetic peptide corresponding to residues Q V Q E S L P E L S N. This immunizing peptide (PEP-303) is available for neutralization and control experiments.
PA1-1755 can be used with blocking peptide PEP-301.
The inter-alpha-trypsin inhibitors (ITI) are a family of structurally related plasma serine protease inhibitors involved in extracellular matrix stabilization and in prevention of tumor metastasis. The ITI family contains multiple proteins made up of a light chain (see MIM 176870) and a variable number of heavy chains (Salier et al., 1987 [PubMed 2446322]; Himmelfarb et al., 2004 [PubMed 14744536]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CYCSP39; HCP39; inter-alpha (globulin) inhibitor, H1 polypeptide; inter-alpha-inhibitor heavy chain 1; Inter-alpha-trypsin inhibitor complex component III; Inter-alpha-trypsin inhibitor heavy chain H1; ITI heavy chain H1; MGC126415; Serum-derived hyaluronan-associated protein; SHAP
Gene Aliases: H1P; IATIH; IGHEP1; Intin1; ITI-HC1; ITIH; Itih-1; ITIH1; SHAP
UniProt ID: (Human) P19827, (Mouse) Q61702
Entrez Gene ID: (Human) 3697, (Mouse) 16424
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support