Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVKVTFNSAL AQKETKKDEP KSGEEALIIP PDAVAVDCKD PDDVVPVGQR RAWCWCMCFG LAFMLAGVIL GGAYLYKYFA LQPDDVYYCG IKYIKDDVIL NEPSADAPAA LYQTIEENIK IFEEEEVEFI SVPVPEFADS DPANIVHDFN KKLTAYLDLN LDKCYVIPLN TSIVMPPRNL LELLINIKAG TYLPQSYLIH EHMVITDRIE NIDHLGFFIY RLCHDKETYK LQRRETIKGI QKREASNCFA IRHFENKFAV ETLICS
Amyloid precursor proteins are processed by beta-secretase and gamma-secretase to produce beta-amyloid peptides which form the characteristic plaques of Alzheimer disease. This gene encodes a transmembrane protein which is processed at the C-terminus by furin or furin-like proteases to produce a small secreted peptide which inhibits the deposition of beta-amyloid. Mutations which result in extension of the C-terminal end of the encoded protein, thereby increasing the size of the secreted peptide, are associated with two neurogenerative diseases, familial British dementia and familial Danish dementia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABri/ADan amyloid peptide; Bri; BRICHOS domain containing 2B; imBRI2; Immature BRI2; Integral membrane protein 2B; OTTHUMP00000018394; Protein E25B; RP11-196P14.1; Transmembrane protein BRI
Gene Aliases: ABRI; BRI; BRI2; BRICD2B; E25B; E3-16; FBD; imBRI2; ITM2B; RDGCA
UniProt ID: (Human) Q9Y287
Entrez Gene ID: (Human) 9445
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support