Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 86%; Dog: 86%; Horse: 86%; Human: 100%; Mouse: 83%; Pig: 86%; Rat: 83%
IZUMO4 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Izumo sperm-egg fusion protein 4; putative SAMP14 binding protein variant 1; putative SAMP14 binding protein variant 2; Sperm 22 kDa protein c113
Gene Aliases: C19orf36; IMAGE:4215339; IZUMO4; UNQ831/PRO1758
UniProt ID: (Human) Q1ZYL8
Entrez Gene ID: (Human) 113177
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support