Product References
Rad52 mediates class-switch DNA recombination to IgD.
Nature communications
Xu Y,Zhou H,Post G,Zan H,Casali P
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Immunohistochemistry
Mon Feb 21 00:00:00 UTC 2022
Nanobodies from camelid mice and llamas neutralize SARS-CoV-2 variants.
Nature
Xu J,Xu K,Jung S,Conte A,Lieberman J,Muecksch F,Lorenzi JCC,Park S,Schmidt F,Wang Z,Huang Y,Luo Y,Nair MS,Wang P,Schulz JE,Tessarollo L,Bylund T,Chuang GY,Olia AS,Stephens T,Teng IT,Tsybovsky Y,Zhou T,Munster V,Ho DD,Hatziioannou T,Bieniasz PD,Nussenzweig MC,Kwong PD,Casellas R
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Thu Jul 01 00:00:00 UTC 2021
Loop extrusion mediates physiological Igh locus contraction for RAG scanning.
Nature
Dai HQ,Hu H,Lou J,Ye AY,Ba Z,Zhang X,Zhang Y,Zhao L,Yoon HS,Chapdelaine-Williams AM,Kyritsis N,Chen H,Johnson K,Lin S,Conte A,Casellas R,Lee CS,Alt FW
11-5790-81 was used in Flow cytometry/Cell sorting to find that the expression of wings apart-like protein homologue (WAPL)10, a cohesin-unloading factor, was low in primary progenitor B cells compared with v-Abl-transformed progenitor B cell lines that lacked contraction and RAG scanning of the VH locus.
Mon Feb 01 00:00:00 UTC 2021
Smc3 dosage regulates B cell transit through germinal centers and restricts their malignant transformation.
Nature immunology
Rivas MA,Meydan C,Chin CR,Challman MF,Kim D,Bhinder B,Kloetgen A,Viny AD,Teater MR,McNally DR,Doane AS,Béguelin W,Fernández MTC,Shen H,Wang X,Levine RL,Chen Z,Tsirigos A,Elemento O,Mason CE,Melnick AM
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2021
The dystonia gene THAP1 controls DNA double-strand break repair choice.
Molecular cell
Shinoda K,Zong D,Callen E,Wu W,Dumitrache LC,Belinky F,Chari R,Wong N,Ishikawa M,Stanlie A,Multhaupt-Buell T,Sharma N,Ozelius L,Ehrlich M,McKinnon PJ,Nussenzweig A
11-5790-81 was used in Flow cytometry/Cell sorting to report that the transcription factors THAP1, YY1, and HCF1 bind directly to the SHLD1 promoter, where they cooperatively maintain the low basal expression of SHLD1, thereby ensuring a proper balance between end protection and resection during double-strand DNA breaks repair.
Thu Jun 17 00:00:00 UTC 2021
A platform-agnostic, function first-based antibody discovery strategy using plasmid-free mammalian expression of antibodies.
mAbs
Zhang R,Prabakaran P,Yu X,Mackness BC,Boudanova E,Hopke J,Sancho J,Saleh J,Cho H,Zhang N,Simonds-Mannes H,Stimple SD,Hoffmann D,Park A,Chowdhury PS,Rao SP
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Wed Jan 19 00:00:00 UTC 2022
The Actin-Disassembly Protein Glia Maturation Factor γ Enhances Actin Remodeling and B Cell Antigen Receptor Signaling at the Immune Synapse.
Frontiers in cell and developmental biology
Deretic N,Bolger-Munro M,Choi K,Abraham L,Gold MR
11-5790-81 was used in Flow Cytometry to suggest that GMFγ cooperates with the Arp2/3 complex to support BCR-induced actin remodeling and amplify BCR signaling at the immune synapse.
Tue Aug 03 00:00:00 UTC 2021
Stearoyl-CoA Desaturase-Mediated Monounsaturated Fatty Acid Availability Supports Humoral Immunity.
Cell reports
Zhou X,Zhu X,Li C,Li Y,Ye Z,Shapiro VS,Copland JA,Hitosugi T,Bernlohr DA,Sun J,Zeng H
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in ELISA
Tue Jan 05 00:00:00 UTC 2021
Primary Immune Responses and Affinity Maturation Are Controlled by IgD.
Frontiers in immunology
Amendt T,Ayoubi OE,Linder AT,Allies G,Young M,Setz CS,Jumaa H
11-5790-81 was used in Immunohistochemistry to suggest that IgD is required for the transition from primary, highly autoreactive, to secondary antigen-specific antibody responses generated by affinity maturation.
Mon Dec 20 00:00:00 UTC 2021
Construction, characterization, and immunization of nanoparticles that display a diverse array of influenza HA trimers.
PloS one
Cohen AA,Yang Z,Gnanapragasam PNP,Ou S,Dam KA,Wang H,Bjorkman PJ
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Fri Oct 15 00:00:00 UTC 2021
Plasmacytoid Dendritic Cells and Type I Interferon Promote Extrafollicular B Cell Responses to Extracellular Self-DNA.
Immunity
Soni C,Perez OA,Voss WN,Pucella JN,Serpas L,Mehl J,Ching KL,Goike J,Georgiou G,Ippolito GC,Sisirak V,Reizis B
11-5790-81 was used in Flow Cytometry to establish extrafollicular B cell differentiation into short-lived AFCs as a key mechanism of anti-DNA autoreactivity and reveal a major contribution of pDCs, endosomal Toll-like receptors (TLRs), and IFN-I to this pathway.
Tue Jun 16 00:00:00 UTC 2020
B Cell Diversification Is Uncoupled from SAP-Mediated Selection Forces in Chronic Germinal Centers within Peyer's Patches.
Cell reports
Biram A,Winter E,Denton AE,Zaretsky I,Dassa B,Bemark M,Linterman MA,Yaari G,Shulman Z
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Tue Feb 11 00:00:00 UTC 2020
Chronic Viral Infection Promotes Efficient Germinal Center B Cell Responses.
Cell reports
Fallet B,Hao Y,Florova M,Cornille K,de Los Aires AV,Girelli Zubani G,Ertuna YI,Greiff V,Menzel U,Hammad K,Merkler D,Reddy ST,Weill JC,Reynaud CA,Pinschewer DD
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Tue Jan 28 00:00:00 UTC 2020
TET enzymes control antibody production and shape the mutational landscape in germinal centre B cells.
The FEBS journal
Schoeler K,Aufschnaiter A,Messner S,Derudder E,Herzog S,Villunger A,Rajewsky K,Labi V
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Sun Sep 01 00:00:00 UTC 2019
Distinct Requirements of CHD4 during B Cell Development and Antibody Response.
Cell reports
Yen WF,Sharma R,Cols M,Lau CM,Chaudhry A,Chowdhury P,Yewdell WT,Vaidyanathan B,Sun A,Coffre M,Pucella JN,Chen CC,Jasin M,Sun JC,Rudensky AY,Koralov SB,Chaudhuri J
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in ELISA
Tue Apr 30 00:00:00 UTC 2019
Anti‑IL‑39 (IL‑23p19/Ebi3) polyclonal antibodies ameliorate autoimmune symptoms in lupus‑like mice.
Molecular medicine reports
Wang X,Zhang Y,Wang Z,Liu X,Zhu G,Han G,Chen G,Hou C,Wang T,Shen B,Li Y,Xiao H,Ma N,Wang R
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in ELISA
Mon Jan 01 00:00:00 UTC 2018
A protocol for generating induced T cells by reprogramming B cells in vivo.
Cell regeneration (London, England)
Weng Q,Hu F,Zhang M,Dong Y,Lv C,Wang Y,Liu X,Wang J
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Sat Sep 01 00:00:00 UTC 2018
PI3Kδ hyper-activation promotes development of B cells that exacerbate Streptococcus pneumoniae infection in an antibody-independent manner.
Nature communications
Stark AK,Chandra A,Chakraborty K,Alam R,Carbonaro V,Clark J,Sriskantharajah S,Bradley G,Richter AG,Banham-Hall E,Clatworthy MR,Nejentsev S,Hamblin JN,Hessel EM,Condliffe AM,Okkenhaug K
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in T-Cell Activation
Thu Aug 09 00:00:00 UTC 2018
CTCF-Binding Elements Mediate Accessibility of RAG Substrates During Chromatin Scanning.
Cell
Jain S,Ba Z,Zhang Y,Dai HQ,Alt FW
11-5790-81 was used in Flow cytometry/Cell sorting to report that CTCF-binding elements (CBEs) immediately downstream of frequently rearranged VH-RSSs increase recombination potential of their associated VH far beyond that provided by RSSs alone.
Thu Jun 28 00:00:00 UTC 2018
TLR4 signals in B lymphocytes are transduced via the B cell antigen receptor and SYK.
The Journal of experimental medicine
Schweighoffer E,Nys J,Vanes L,Smithers N,Tybulewicz VLJ
11-5790 was used in Flow cytometry/Cell sorting to demonstrate two distinct signalling pathways for TLR4, one via the BCR and the other through MYD88.
Mon May 01 00:00:00 UTC 2017
Conversion of T cells to B cells by inactivation of polycomb-mediated epigenetic suppression of the B-lineage program.
Genes & development
Ikawa T,Masuda K,Endo TA,Endo M,Isono K,Koseki Y,Nakagawa R,Kometani K,Takano J,Agata Y,Katsura Y,Kurosaki T,Vidal M,Koseki H,Kawamoto H
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in ELISA
Tue Nov 15 00:00:00 UTC 2016
Conversion of T cells to B cells by inactivation of polycomb-mediated epigenetic suppression of the B-lineage program.
Genes & development
Ikawa T,Masuda K,Endo TA,Endo M,Isono K,Koseki Y,Nakagawa R,Kometani K,Takano J,Agata Y,Katsura Y,Kurosaki T,Vidal M,Koseki H,Kawamoto H
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in ELISA
Tue Nov 15 00:00:00 UTC 2016
Ligase I and ligase III mediate the DNA double-strand break ligation in alternative end-joining.
Proceedings of the National Academy of Sciences of the United States of America
Lu G,Duan J,Shu S,Wang X,Gao L,Guo J,Zhang Y
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Tue Feb 02 00:00:00 UTC 2016
Mast cell depletion in the preclinical phase of collagen-induced arthritis reduces clinical outcome by lowering the inflammatory cytokine profile.
Arthritis research & therapy
van der Velden D,Lagraauw HM,Wezel A,Launay P,Kuiper J,Huizinga TW,Toes RE,Bot I,Stoop JN
11-5790 was used in Flow cytometry/Cell sorting to suggest that mast cells may play a role in the regulation of the adaptive immune response during the development of arthritis.
Mon Jun 13 00:00:00 UTC 2016
Global Analysis of O-GlcNAc Glycoproteins in Activated Human T Cells.
Journal of immunology (Baltimore, Md. : 1950)
Lund PJ,Elias JE,Davis MM
11-5790 was used in Flow cytometry/Cell sorting to provide a global analysis of O-GlcNAc dynamics during T cell activation and the first characterization.
Sat Oct 15 00:00:00 UTC 2016
Editing of mouse and human immunoglobulin genes by CRISPR-Cas9 system.
Nature communications
Cheong TC,Compagno M,Chiarle R
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Wed Mar 09 00:00:00 UTC 2016
BCL11A enhancer dissection by Cas9-mediated in situ saturating mutagenesis.
Nature
Canver MC,Smith EC,Sher F,Pinello L,Sanjana NE,Shalem O,Chen DD,Schupp PG,Vinjamur DS,Garcia SP,Luc S,Kurita R,Nakamura Y,Fujiwara Y,Maeda T,Yuan GC,Zhang F,Orkin SH,Bauer DE
11-5790 was used in Flow cytometry/Cell sorting to develop pooled CRISPR-Cas9 guide RNA libraries to perform in situ saturating mutagenesis of human and mouse enhancers.
Thu Nov 12 00:00:00 UTC 2015
B cell autophagy mediates TLR7-dependent autoimmunity and inflammation.
Autophagy
Weindel CG,Richey LJ,Bolland S,Mehta AJ,Kearney JF,Huber BT
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Mon Apr 18 00:00:00 UTC 2016
B cell autophagy mediates TLR7-dependent autoimmunity and inflammation.
Autophagy
Weindel CG,Richey LJ,Bolland S,Mehta AJ,Kearney JF,Huber BT
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Mon Apr 18 00:00:00 UTC 2016
B cell autophagy mediates TLR7-dependent autoimmunity and inflammation.
Autophagy
Weindel CG,Richey LJ,Bolland S,Mehta AJ,Kearney JF,Huber BT
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Mon Apr 18 00:00:00 UTC 2016
Mitochondrial ATP transporter Ant2 depletion impairs erythropoiesis and B lymphopoiesis.
Cell death and differentiation
Cho J,Seo J,Lim CH,Yang L,Shiratsuchi T,Lee MH,Chowdhury RR,Kasahara H,Kim JS,Oh SP,Lee YJ,Terada N
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2015
Splenic extrafollicular reactions and BM plasma cells sustain IgM response associated with hypercholesterolemia.
European journal of immunology
Khoo LH,Thiam CH,Soh SY,Angeli V
11-5790 was used in Flow cytometry/Cell sorting to elucidate the mechanism underlying the elevated IgM response observed in hypercholesterolaemia.
Fri May 01 00:00:00 UTC 2015
Natural IgM is produced by CD5- plasma cells that occupy a distinct survival niche in bone marrow.
Journal of immunology (Baltimore, Md. : 1950)
Reynolds AE,Kuraoka M,Kelsoe G
11-5790 was used in Flow cytometry/Cell sorting to investigate the origin of natural IgM, showing that it is produced by CD5- plasma cells that occupy a distinct survival niche in bone marrow.
Thu Jan 01 00:00:00 UTC 2015
Core fucosylation of IgG B cell receptor is required for antigen recognition and antibody production.
Journal of immunology (Baltimore, Md. : 1950)
Li W,Yu R,Ma B,Yang Y,Jiao X,Liu Y,Cao H,Dong W,Liu L,Ma K,Fukuda T,Liu Q,Ma T,Wang Z,Gu J,Zhang J,Taniguchi N
11-5790 was used in Flow cytometry/Cell sorting to investigate core fucosylation of IgG B cell receptor's role in antigen recognition and antibody production.
Sun Mar 15 00:00:00 UTC 2015
VprBP Is Required for Efficient Editing and Selection of Igκ+ B Cells, but Is Dispensable for Igλ+ and Marginal Zone B Cell Maturation and Selection.
Journal of immunology (Baltimore, Md. : 1950)
Palmer VL,Aziz-Seible R,Kassmeier MD,Rothermund M,Perry GA,Swanson PC
11-5790 was used in Flow cytometry/Cell sorting to determine the contribution of the protein, VprBP, to B cell development and selection.
Sat Aug 15 00:00:00 UTC 2015
The proximal J kappa germline-transcript promoter facilitates receptor editing through control of ordered recombination.
PloS one
Vettermann C,Timblin GA,Lim V,Lai EC,Schlissel MS
11-5790 was used in Flow cytometry/Cell sorting to identify the proximal GT promoter as the first cis-element known to regulate the internal order of Jκ rearrangements.
Tue Jan 26 00:00:00 UTC 2016
Induced Developmental Arrest of Early Hematopoietic Progenitors Leads to the Generation of Leukocyte Stem Cells.
Stem cell reports
Ikawa T,Masuda K,Huijskens MJAJ,Satoh R,Kakugawa K,Agata Y,Miyai T,Germeraad WTV,Katsura Y,Kawamoto H
11-5790 was used in Flow cytometry/Cell sorting to show that a simple block of cell differentiation is sufficient to induce and maintain stem cells.
Tue Nov 10 00:00:00 UTC 2015
Elucidation of the enigmatic IgD class-switch recombination via germline deletion of the IgH 3' regulatory region.
The Journal of experimental medicine
Rouaud P,Saintamand A,Saad F,Carrion C,Lecardeur S,Cogné M,Denizot Y
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Immunohistochemistry
Mon May 05 00:00:00 UTC 2014
Elucidation of the enigmatic IgD class-switch recombination via germline deletion of the IgH 3' regulatory region.
The Journal of experimental medicine
Rouaud P,Saintamand A,Saad F,Carrion C,Lecardeur S,Cogné M,Denizot Y
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Immunohistochemistry
Mon May 05 00:00:00 UTC 2014
B cell activation involves nanoscale receptor reorganizations and inside-out signaling by Syk.
eLife
Kläsener K,Maity PC,Hobeika E,Yang J,Reth M
11-5790 was used in Flow cytometry/Cell sorting to provide direct evidence for the opening of B cell antigen receptor dimers during B cell activation.
Tue Jun 24 00:00:00 UTC 2014
Ectopic lymphoid tissue alters the chemokine gradient, increases lymphocyte retention and exacerbates murine ileitis.
Gut
McNamee EN,Masterson JC,Jedlicka P,Collins CB,Williams IR,Rivera-Nieves J
11-5790 was used in Immunofluorescence to study the contribution of the CCR7/CCL19/CCL21 chemokine axis during the development of tertiary lymphoid tissue and its role in disease.
Tue Jan 01 00:00:00 UTC 2013
A component of the mir-17-92 polycistronic oncomir promotes oncogene-dependent apoptosis.
eLife
Olive V,Sabio E,Bennett MJ,De Jong CS,Biton A,McGann JC,Greaney SK,Sodir NM,Zhou AY,Balakrishnan A,Foth M,Luftig MA,Goga A,Speed TP,Xuan Z,Evan GI,Wan Y,Minella AC,He L
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Tue Oct 15 00:00:00 UTC 2013
A mouse model for inducible overexpression of Prdm14 results in rapid-onset and highly penetrant T-cell acute lymphoblastic leukemia (T-ALL).
Disease models & mechanisms
Carofino BL,Ayanga B,Justice MJ
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Fri Nov 01 00:00:00 UTC 2013
B cell activating factor inhibition impairs bacterial immunity by reducing T cell-independent IgM secretion.
Infection and immunity
Jones DD,Jones M,DeIulio GA,Racine R,MacNamara KC,Winslow GM
11-5790 was used in Flow cytometry/Cell sorting to determine that BAFF signalling can directly regulate secretory processes.
Sun Dec 01 00:00:00 UTC 2013
IKKα-mediated signaling circuitry regulates early B lymphopoiesis during hematopoiesis.
Blood
Balkhi MY,Willette-Brown J,Zhu F,Chen Z,Liu S,Guttridge DC,Karin M,Hu Y
11-5790-81 was used in Flow Cytometry to demonstrate that IKKα regulates B-lineage commitment via combined canonical and noncanonical NF-κB transcriptional activities to target Pax5 expression during hematopoiesis.
Thu Jun 07 00:00:00 UTC 2012
Nucleoside salvage pathway kinases regulate hematopoiesis by linking nucleotide metabolism with replication stress.
The Journal of experimental medicine
Austin WR,Armijo AL,Campbell DO,Singh AS,Hsieh T,Nathanson D,Herschman HR,Phelps ME,Witte ON,Czernin J,Radu CG
11-5790 was used in Flow cytometry/Cell sorting to investigate a functional link between nucleotide deficiency, replication stress, and two enzymes of the nucleoside salvage pathway: deoxycytidine kinase and thymidine kinase.
Mon Nov 19 00:00:00 UTC 2012
Critical role of B cell lymphoma 10 in BAFF-regulated NF-κB activation and survival of anergic B cells.
Journal of immunology (Baltimore, Md. : 1950)
Yu M,Chen Y,He Y,Podd A,Fu G,Wright JA,Kleiman E,Khan WN,Wen R,Wang D
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Sat Dec 01 00:00:00 UTC 2012
Nuclear export of the NF-κB inhibitor IκBα is required for proper B cell and secondary lymphoid tissue formation.
Immunity
Wuerzberger-Davis SM,Chen Y,Yang DT,Kearns JD,Bates PW,Lynch C,Ladell NC,Yu M,Podd A,Zeng H,Huang TT,Wen R,Hoffmann A,Wang D,Miyamoto S
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Fri Feb 25 00:00:00 UTC 2011
Nuclear export of the NF-κB inhibitor IκBα is required for proper B cell and secondary lymphoid tissue formation.
Immunity
Wuerzberger-Davis SM,Chen Y,Yang DT,Kearns JD,Bates PW,Lynch C,Ladell NC,Yu M,Podd A,Zeng H,Huang TT,Wen R,Hoffmann A,Wang D,Miyamoto S
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Fri Feb 25 00:00:00 UTC 2011
IgM production by bone marrow plasmablasts contributes to long-term protection against intracellular bacterial infection.
Journal of immunology (Baltimore, Md. : 1950)
Racine R,McLaughlin M,Jones DD,Wittmer ST,MacNamara KC,Woodland DL,Winslow GM
11-5790 was used in Flow cytometry/Cell sorting to identify a cell population responsible for IgM production in the bone marrow that is maintained in the absence of chronic infection.
Sat Jan 15 00:00:00 UTC 2011
The actin-bundling protein L-plastin is essential for marginal zone B cell development.
Journal of immunology (Baltimore, Md. : 1950)
Todd EM,Deady LE,Morley SC
11-5790 was used in Flow cytometry/Cell sorting to study the role of the actin-bundling protein L-plastin in B cell chemotaxis.
Thu Sep 15 00:00:00 UTC 2011
HVCN1 modulates BCR signal strength via regulation of BCR-dependent generation of reactive oxygen species.
Nature immunology
Capasso M,Bhamrah MK,Henley T,Boyd RS,Langlais C,Cain K,Dinsdale D,Pulford K,Khan M,Musset B,Cherny VV,Morgan D,Gascoyne RD,Vigorito E,DeCoursey TE,MacLennan IC,Dyer MJ
11-5790 was used in Flow cytometry/Cell sorting to investigate how HVCN1 modulates BCR signal strength through regulation of BCR-dependent generation of reactive oxygen species.
Mon Mar 01 00:00:00 UTC 2010
Innate retroviral restriction by Apobec3 promotes antibody affinity maturation in vivo.
Journal of immunology (Baltimore, Md. : 1950)
Santiago ML,Benitez RL,Montano M,Hasenkrug KJ,Greene WC
11-5790 was used in Flow cytometry/Cell sorting to propose that enabling Apobec3 activity during acute infection with human pathogenic retroviruses may similarly facilitate stronger virus-specific neutralizing Ab responses.
Thu Jul 15 00:00:00 UTC 2010
A critical role of TAK1 in B-cell receptor-mediated nuclear factor kappaB activation.
Blood
Schuman J,Chen Y,Podd A,Yu M,Liu HH,Wen R,Chen ZJ,Wang D
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Thu May 07 00:00:00 UTC 2009
A critical role of TAK1 in B-cell receptor-mediated nuclear factor kappaB activation.
Blood
Schuman J,Chen Y,Podd A,Yu M,Liu HH,Wen R,Chen ZJ,Wang D
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Thu May 07 00:00:00 UTC 2009
miR-19 is a key oncogenic component of mir-17-92.
Genes & development
Olive V,Bennett MJ,Walker JC,Ma C,Jiang I,Cordon-Cardo C,Li QJ,Lowe SW,Hannon GJ,He L
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Tue Dec 15 00:00:00 UTC 2009
Phospholipase Cgamma2 contributes to light-chain gene activation and receptor editing.
Molecular and cellular biology
Bai L,Chen Y,He Y,Dai X,Lin X,Wen R,Wang D
Published figure using IgM monoclonal antibody (Product # 11-5790-81) in Flow Cytometry
Sat Sep 01 00:00:00 UTC 2007