Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Suggested positive control: HeLa lysate, antigen standard for IPO11 (transient overexpression lysate).
IPO11 functions in nuclear protein import as nuclear transport receptor. It serves as receptor for nuclear localization signals (NLS) in cargo substrates and is thought to mediate docking of the importin/substrate complex to the nuclear pore complex (NPC) through binding to nucleoporin and the complex is subsequently translocated through the pore by an energy requiring, Ran-dependent mechanism. At the nucleoplasmic side of the NPC, Ran binds to the importin, the importin/substrate complex dissociates and importin is re-exported from the nucleus to the cytoplasm where GTP hydrolysis releases Ran. The directionality of nuclear import is thought to be conferred by an asymmetric distribution of the GTP-and GDP-bound forms of Ran between the cytoplasm and nucleus. It mediates the nuclear import of UBE2E3, and of RPL12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Imp11; Importin-11; Ran binding protein 11; Ran-binding protein 11; RanBP11
Gene Aliases: 1700081H05Rik; 2510001A17Rik; AI314624; AW555235; E330021B14Rik; IPO11; RANBP11
UniProt ID: (Human) Q9UI26, (Mouse) Q8K2V6
Entrez Gene ID: (Dog) 478073, (Bovine) 538236, (Human) 51194, (Rat) 310056, (Mouse) 76582
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support