Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total IPO13 protein.
This gene encodes a member of the importin-beta family of nuclear transport proteins. The encoded protein mediates the import of specific cargo proteins from the cytoplasm to the nucleus and is dependent on the Ras-related nuclear protein-GTPase system. The encoded protein is also involved in nuclear export of the eukaryotic translation initiation factor 1A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Imp13; Importin-13; Kap13; karyopherin 13; Karyopherin-13; late gestation lung 2; Ran binding protein 13; Ran-binding protein 13; RanBP13; RP11-7O11.4
Gene Aliases: IMP13; IPO13; KAP13; KIAA0724; LGL2; RANBP13
UniProt ID: (Human) O94829
Entrez Gene ID: (Human) 9670
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support