Invitrogen
FIGURE: 1 / 4
Recommended positive controls: Influenza virus (A/WSN/1933(H1N1))-infected DF-1 cell lysates, Influenza virus-infected monkey vero cells.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Influenza A viruses are single stranded, negative sense RNA viruses. Two distinct non-structural proteins NS1 and NS2 are translated from one of the eight RNA segments by using different reading frames.The influenza A virus NS2 is located in the nucleus and the cytoplasm. NS2 is essential for nuclear export of influenza virus ribonucleoprotein (RNP) complexes.