Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Activin A mutations are associated with fibrodysplasia ossificans progressive.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activin A; Activin beta-A chain; EDF; erythroid differentiation factor; Erythroid differentiation protein; follicle-stimulating hormone-releasing protein; FSH-releasing protein; Inhibin beta A chain; inhibin, beta A (activin A, activin AB alpha polypeptide); Inhibin, beta-1
Gene Aliases: EDF; FRP; INHBA
UniProt ID: (Human) P08476, (Rat) P18331, (Mouse) Q04998
Entrez Gene ID: (Human) 3624, (Rat) 29200, (Mouse) 16323
Molecular Function: growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support