Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha v integrin; antigen identified by monoclonal antibody L230; CD51; Dnmt3l-ps1 pseudogene; integrin alpha V; Integrin alpha-V; integrin alphaVbeta3; integrin beta 6; Integrin beta-6; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); integrin, beta 6; Vitronectin receptor; vitronectin receptor alpha polypeptide (VNRA); Vitronectin receptor subunit alpha
Gene Aliases: 1110004F14Rik; 2210409C20Rik; 2610028E01Rik; 4831415H04Rik; AI1H; CD51; D430040G12Rik; ITGAV; ITGB6; MSK8; VNRA; VTNR
UniProt ID: (Human) P06756, (Human) P18564, (Mouse) P43406, (Rat) Q6AYF4, (Mouse) Q9Z0T9
Entrez Gene ID: (Human) 3685, (Human) 3694, (Rat) 296456, (Mouse) 16410, (Rat) 311061, (Mouse) 16420
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support